Skip to Content
Merck
All Photos(3)

Key Documents

P0056

Sigma-Aldrich

Anti-POU5F1 (Oct4) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-OCT3, Anti-OCT4, Anti-OTF3, Anti-OTF4, Anti-Octamer-binding transcription factor 3, Anti-Pou domain, Class 5, Transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using NT2 cell lysate
indirect immunofluorescence: 2-5 μg/mL using paraformaldhyde-fixed F9 cells
western blot: 1-2 μg/mL using NT2 cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)
mouse ... Pou5f1(18999)
rat ... Pou5f1(294562)

General description

POU class 5 homeobox 1 (POU5F1) recognizes defined octamer DNA element by a shared, conserved DNA-binding domain, namely the POU domain.
POU class 5 homeobox 1 (POU5F1), also known as octamer-binding transcription factor-4 (OCT-4), is expressed in embryonic stem cells and germ cells. It belongs to the POU family of transcription factors. Three splice variants of the protein (OCT4A, OCT4B and OCT4B1) have been identified. The gene encoding this transcription factor is localized on human chromosome 6p21.33.

Immunogen

synthetic peptide corresponding to amino acids 19-34 of human POU5F1, conjugated to KLH. The corresponding sequence in rat is 81% homologous.

Application

Anti-POU5F1 (Oct4) antibody produced in rabbit has been used in Western Blotting.

Biochem/physiol Actions

POU class 5 homeobox 1 (POU5F1) may be a tumor initiating stem-like cell (TSC)-related marker. It has a role in stem cell pluripotency and self-renewal, and also modulates differentiation. The protein has been shown to be expressed in several cancers. POU5F1 is also vital for the induced pluripotent stem cell (iPSC) process.
POU5F1 or Oct4/Oct3 (Octamer-binding transcription factor), functions as an early developmental control gene. It has been recognized as fundamental in the maintenance of pluripotency in embryonic stem cells and in primordial germ cells. It is down regulated in all differentiated somatic cell types both in vivo and in vitro. This is being accomplished through increased DNA methylation and structural changes involving the immediate upstream regulatory region. POU5F1 serves as a marker for germ cell tumors (GCTs) eliciting pluripotentiality, especially including embryonal, seminoma/ dysgerminoma/ germinoma.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of POU5F1 (OCT4) gene and its promoter in buffalo ESC-like cells identifies multiple transcription start sites and expression of four pseudogenes
Natwar Singh
A Comprehensive Guide to the Hazardous Properties of Chemical Substances (2012)
OCT4: biological functions and clinical applications as a marker of germ cell neoplasia
Cheng L, et al.
The Journal of Pathology, 211(1), 1-9 (2007)
The evolution of class V POU domain transcription factors in vertebrates and their characterisation in a marsupial
Frankenberg S, et al.
Developmental Biology, 337, 162-170 (2010)
Hepatitis C virus core protein regulates OCT4 expression and promotes cell cycle progression in hepatocellular carcinoma
Jia-Jia Zhou
Oncology Reports (2016)
Huiping Tan et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 3644-3652 (2018-06-01)
BACKGROUND Recent studies have shown that increased mobilization of Long Interspersed Nuclear Elements-1 (L1) can promote the pathophysiology of multiple neurological diseases. However, its role in Huntington's disease (HD) remains unknown. MATERIAL AND METHODS R6/2 mice - a common mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service