Skip to Content
Merck
All Photos(1)

Key Documents

O9014

Sigma-Aldrich

N-(3-Oxodecanoyl)-L-homoserine lactone

≥98% (TLC)

Synonym(s):

3-Oxo-N-[(3s)-2-Oxotetrahydrofuran-3-Yl]decanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23NO4
Molecular Weight:
269.34
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

N-(3-Oxodecanoyl)-L-homoserine lactone,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)CC(=O)N[C@H]1CCOC1=O

InChI

1S/C14H23NO4/c1-2-3-4-5-6-7-11(16)10-13(17)15-12-8-9-19-14(12)18/h12H,2-10H2,1H3,(H,15,17)/t12-/m0/s1

InChI key

KYGIKEQVUKTKRR-LBPRGKRZSA-N

General description

N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL) is among belongs to the group of homoserine lactones that includes N-octanoyl-homoserine lactone (N-C8-HSL), N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL), N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL, N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone.

Application

N-(3-Oxodecanoyl)-L-homoserine lactone (3-Oxo-C10-HSL) may be used to study the signaling involved in the regulation of bacterial quorum sensing.

Biochem/physiol Actions

Acyl homoserine lactone used as an autoinducer of quorum signaling by Pseudomonas putida, Yersinia enterocolitica, and other Gram-negative bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John F Teiber et al.
Methods in molecular biology (Clifton, N.J.), 692, 291-298 (2010-10-30)
Mammalian paraoxonases (PONs) are a unique, highly conserved family of calcium-dependent esterases consisting of PON1, PON2, and PON3. The PONs can hydrolyze the lactone ring of a range of N-acyl-L: -homoserine lactone (AHL) quorum sensing signaling molecules, rendering them inactive.
Rumana Rashid et al.
Microbes and environments, 26(2), 144-148 (2011-04-20)
N-Acylhomoserine lactones (AHLs) are used as quorum-sensing signaling molecules by many Gram-negative bacteria. Here, 413 bacterial strains were obtained from the roots of potato plants and screened for AHL-degrading bacteria using Chromobacterium violaceum reporter strains. Sixty one isolates degraded N-hexanoyl-L-homoserine
Manabu Horikawa et al.
Bioorganic & medicinal chemistry letters, 16(8), 2130-2133 (2006-02-08)
The synthesis of the analogs of N-3-oxododecanoyl-L-homoserine lactone (1) and their structure-activity relationship for the apoptotic induction in macrophages, P388D1 cells, are described. It was revealed that the position of the oxo group in the acyl side chain in addition
Mair E A Churchill et al.
Methods in molecular biology (Clifton, N.J.), 692, 159-171 (2010-10-30)
Quorum sensing plays a central role in regulating many community-derived symbiotic and pathogenic relationships of bacteria, and as such has attracted much attention in recent years. Acyl-homoserine lactones (AHLs) are important signaling molecules in the quorum sensing gene-regulatory processes found
Bijay K Khajanchi et al.
Infection and immunity, 79(7), 2646-2657 (2011-05-04)
Aeromonas hydrophila leads to both intestinal and extraintestinal infections in animals and humans, and the underlying mechanisms leading to mortality are largely unknown. By using a septicemic mouse model of infection, we showed that animals challenged with A. hydrophila die

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service