Skip to Content
Merck
All Photos(4)

Key Documents

O6004

Sigma-Aldrich

Octyl β-D-1-thioglucopyranoside

≥98.0% (GC)

Synonym(s):

Octyl thioglucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O5S
CAS Number:
Molecular Weight:
308.43
Beilstein:
4182783
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

description

non-ionic

Assay

≥98.0% (GC)

form

crystalline

optical activity

[α]25/D -51 to -49 °, c = 1% (w/v) in ethanol

mol wt

308.43 g/mol

CMC

9 mM (20-25°C)

storage temp.

−20°C

SMILES string

CCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O5S/c1-2-3-4-5-6-7-8-20-14-13(18)12(17)11(16)10(9-15)19-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14+/m1/s1

InChI key

CGVLVOOFCGWBCS-RGDJUOJXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octyl β-D-1-thioglucopyranoside can be used as a non-ionic detergent for solubilization and reconstitution of membrane proteins. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

K H Weisgraber et al.
Journal of molecular biology, 236(1), 382-384 (1994-02-11)
Human apolipoprotein C-I, the smallest of the plasma apolipoproteins, is a component of several classes of plasma lipoproteins. Two crystalline forms of this protein, both suitable for high resolution X-ray diffraction analysis, have been obtained from mixtures of 2-methyl-2,4-pentanediol, sodium
M Yoshida et al.
European journal of biochemistry, 222(3), 1055-1061 (1994-06-15)
Dystrophin is purified as a complex with several proteins from the digitonin-solubilized muscle cell membrane. Most of dystrophin-associated proteins (DAPs) are assumed to form a large oligomeric transmembranous glycoprotein complex on the sarcolemma and link dystrophin with a basement membrane
Sanghamitra Mukhopadhyay et al.
Molecular & cellular proteomics : MCP, 5(12), 2311-2318 (2006-08-22)
Chlamydia pneumoniae is an important human respiratory pathogen that is responsible for an estimated 10% of community-acquired pneumonia and 5% of bronchitis and sinusitis cases. We examined changes in global protein expression profiles associated with the redifferentiation of reticulate body
M Chami et al.
Journal of structural biology, 133(1), 64-74 (2001-05-18)
A great interest exists in producing and/or improving two-dimensional (2D) crystals of membrane proteins amenable to structural analysis by electron crystallography. Here we report on the use of the detergent n-octyl beta-d-thioglucopyranoside in 2D crystallization trials of membrane proteins with
Praveen Konidala et al.
Journal of molecular graphics & modelling, 25(1), 77-86 (2006-01-03)
n-Octyl-beta-D-glucopyranoside (OG) is a non-ionic glycolipid, which is used widely in biotechnical and biochemical applications. All-atom molecular dynamics simulations from two different initial coordinates and velocities in explicit solvent have been performed to characterize the structural behaviour of an OG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service