Skip to Content
Merck
All Photos(1)

Key Documents

O4878

Sigma-Aldrich

Oxaloacetate Decarboxylase from Pseudomonas sp.

lyophilized powder, ≥100 units/mg solid

Synonym(s):

OAD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥100 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Oxaloacetate decarboxylase or OAD functions as a Na pump in anaerobic bacteria. It is a membrane protein consisting of three subunits, α, β and γ with the α subunit containing the carboxylase activity.

Application

Oxaloacetate Decarboxylase from Pseudomonas sp. has been used in the digestion of the low molecular weight (LMW) human milk fraction (5kF fraction) and as a positive control for deciphering C. thermocellum oxaloacetate decarboxylase activity.
Oxaloacetate decarboxylase has been used in a study to assess turnover and accessibility of a reentrant loop of the Na(+)-glutamate transporter GltS. It has also been used in a study to investigate fermentation and metabolic characteristics of Gluconacetobacter oboediens for different carbon sources.

Biochem/physiol Actions

Oxaloacetate Decarboxylase catalyzes the decarboxylation of oxaloacetate and requires manganese and magnesium for its activity. It is associated with a wide vareity of Gram-negative bacteria.

Unit Definition

One unit will convert 1.0 μmole of oxalacetate to pyruvate and CO2 per min at pH 8.0 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pius Dahinden et al.
The FEBS journal, 272(3), 846-855 (2005-01-27)
The oxaloacetate decarboxylase Na+ pumps OAD-1 and OAD-2 of Vibrio cholerae are composed of a peripheral alpha-subunit associated with two integral membrane-bound subunits (beta and gamma). The alpha-subunit contains the carboxyltransferase domain in its N-terminal portion and the biotin-binding domain
Monica Balsera et al.
The Journal of biological chemistry, 286(11), 9457-9467 (2011-01-07)
The oxaloacetate decarboxylase primary Na(+) pump (OAD) is an essential membrane protein complex that functions in the citrate fermentation pathway of some pathogenic bacteria under anaerobic conditions. OAD contains three different subunits: Oad-α, a biotinylated extrinsic protein that catalyzes the
Tomas Krupnik et al.
Molecular membrane biology, 28(7-8), 462-472 (2011-10-15)
GltS of Escherichia coli is a secondary transporter that catalyzes Na+-glutamate symport. The structural model of GltS shows two homologous domains with inverted membrane topology that are connected by a central loop that resides in the cytoplasm. Each domain contains
Pablo D Sender et al.
FEBS letters, 570(1-3), 217-222 (2004-07-15)
The citM gene from Lactococcus lactis CRL264 was demonstrated to encode for an oxaloacetate decarboxylase. The enzyme exhibits high levels of similarity to malic enzymes (MEs) from other organisms. CitM was expressed in Escherichia coli, purified and its oxaloacetate decarboxylase
Oxalacetic carboxylase of Micrococcus lysodeikticus.
Herbert, D.
Methods in Enzymology, 1, 753-757 (1955)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service