Skip to Content
Merck
All Photos(3)

Key Documents

O3129

Sigma-Aldrich

N-Octanoyl-N-methylglucamine

≥97% (GC)

Synonym(s):

N-(D-Glucityl)-N-methyloctanamide, N-Methyl-N-octanoyl-D-glucamine, MEGA-8, OMEGA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H31NO6
CAS Number:
Molecular Weight:
321.41
Beilstein:
5816747
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

Assay

≥97% (GC)

form

powder

mol wt

321.41 g/mol

color

white

CMC

58 mM (20-25°C)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

CCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C15H31NO6/c1-3-4-5-6-7-8-13(20)16(2)9-11(18)14(21)15(22)12(19)10-17/h11-12,14-15,17-19,21-22H,3-10H2,1-2H3/t11-,12+,14+,15+/m0/s1

InChI key

SBWGZAXBCCNRTM-CTHBEMJXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Octanoyl-N-methylglucamine has been used in a study to assess disordered the N-terminus of PKA. It has also been used in a study to investigate the usefulness of sugar surfactants as solubilizing agents in parenteral formulations.
Non-ionic detergent used for the solubilization of proteins

Biochem/physiol Actions

Water soluble detergent with high solubilising power and non-denaturating properties. Readily removed by dialysis.; Does not interfere with the monitoring of proteins at 280 nm.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sverre Bang Småge et al.
PloS one, 12(11), e0187476-e0187476 (2017-11-03)
Tenacibaculosis is an increasing problem in the Norwegian Atlantic salmon aquaculture industry causing significant economic losses. In September 2015, two separate outbreaks of suspected tenacibaculosis occurred at two Atlantic salmon farms in Finnmark County in Northern Norway. The events resulted
Rerngchai Arayanarakool et al.
Micromachines, 9(10) (2018-11-15)
This work presents the simple and rapid fabrication of a polymer-based microfluidic prototype manufactured by rolling up thin films of polymer. The thin films were fabricated via a casting method and rolled up around a center core with the aid
Guangtao Zhu et al.
Cell, 172(1-2), 249-261 (2018-01-13)
Humans heavily rely on dozens of domesticated plant species that have been further improved through intensive breeding. To evaluate how breeding changed the tomato fruit metabolome, we have generated and analyzed a dataset encompassing genomes, transcriptomes, and metabolomes from hundreds
Yange Xu et al.
BMC genomics, 18(1), 727-727 (2017-09-15)
Suaeda salsa (S. salsa) is a euhalophyte with high economic value. S. salsa can produce dimorphic seeds. Brown seeds are more salt tolerant, can germinate quickly and maintain the fitness of the species under high saline conditions. Black seeds are
Assessment of the Diversity of
Jun Chen et al.
Frontiers in microbiology, 8, 1748-1748 (2017-10-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service