Skip to Content
Merck
All Photos(1)

Key Documents

O0257

Sigma-Aldrich

Olvanil

powder

Synonym(s):

(N-Vanillyl)-9-oleamide, N-Vannilyloleoylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H43NO3
CAS Number:
Molecular Weight:
417.62
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

white to off-white

solubility

H2O: <0.2 mg/mL
DMSO: 20 mg/mL

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)NCc1ccc(O)c(OC)c1

InChI

1S/C26H43NO3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-26(29)27-22-23-19-20-24(28)25(21-23)30-2/h10-11,19-21,28H,3-9,12-18,22H2,1-2H3,(H,27,29)/b11-10-

InChI key

OPZKBPQVWDSATI-KHPPLWFESA-N

Gene Information

human ... TRPV1(7442)

Application

Olvanil has been used as a vanilloid (VR1) receptors agonist in rat VR1-transfected human embryonic kidney cells (rVR1-HEK) and dorsal root ganglion (DRG) cells to test its effect on calcium levels.

Biochem/physiol Actions

Olvanil is a vanilloid receptor agonist. It is an analog of capsaicin and is non-pungent. It is an orally active analgesic that alleviates pain by desensitizing nociceptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamaki Hayase
BMC pharmacology, 11, 6-6 (2011-07-20)
The contributions of brain cannabinoid (CB) receptors, typically CB1 (CB type 1) receptors, to the behavioral effects of nicotine (NC) have been reported to involve brain transient receptor potential vanilloid 1 (TRPV1) receptors, and the activation of candidate endogenous TRPV1
Edmundo Castillo et al.
Biochemical and biophysical research communications, 356(2), 424-430 (2007-03-17)
Capsaicin (Cap) and its analogs (CAPanalogs) have diverse effects in sensory neurons including analgesia, implying they modulate other cellular targets besides the TRPV1 Cap receptor. Since Cap and CAPanalogs are not largely available and their chemical synthesis is cumbersome, they
Vincenzo Micale et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 34(3), 593-606 (2008-06-27)
The endocannabinoid-inactivating enzyme, fatty acid amide hydrolase (FAAH), and the transient receptor potential vanilloid type-1 (TRPV1) channel are new targets for the development of anxiolytic drugs. We studied the effect on anxiety-like behavior in the elevated plus maze of a
Min-Goo Lee et al.
British journal of pharmacology, 146(4), 596-603 (2005-08-02)
Certain fatty acid amides such as anandamide (AEA) and olvanil are agonists for the transient receptor potential, vanilloid-1 (TRPV1) receptor, but have been found to activate TRPV1-containing C-fibers in some tissues but not others. We used extracellular recording and whole-cell
M Beltramo et al.
European journal of pharmacology, 364(1), 75-78 (1999-01-27)
The structural similarities between the anandamide transport inhibitor N-(4-hydroxyphenyl)-arachidonylamide (AM404) and the synthetic vanilloid agonist olvanil [(N-vanillyl)-9-oleamide], prompted us to investigate the possibility that olvanil may interfere with anandamide transport. The intracellular accumulation of [3H]anandamide by human astrocytoma cells was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service