Skip to Content
Merck
All Photos(3)

Key Documents

NA2110

Sigma-Aldrich

GenElute Bacterial Genomic DNA Kits

sufficient for 70 purifications

Synonym(s):

Bacterial Genomic DNA, bacterial DNA prep, bacterial DNA purification kit, bacterial gDNA extraction kit, bacterial gDNA isolation kit, bacterial gDNA purification kit, bacterial genomic DNA extraction kit, bacterial genomic DNA isolation kit, bacterial genomic DNA purification kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55
Pricing and availability is not currently available.

usage

sufficient for 70 purifications

Quality Level

technique(s)

DNA purification: suitable

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Bacterial Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from gram-negative bacteria. For most gram-positive bacteria, the kit must be used in conjunction with the optional lysozyme (L4919) to effectively lyse the thick peptidoglycan cell walls. A Gram-Positive Lysis Solution is provided as a diluent for preparing the lysozyme stock solutions.

The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified bacterial genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • cloning

Features and Benefits

  • Starting material: Up to 1.5 mL of culture
  • Expected yield: Up to 20 μg
  • Elution volume: 400 μl
  • Time required: 70 - 120 min
  • A260/A280 ratio: 1.6 - 1.9
  • No phenol, chloroform, or ethanol precipitation required
  • Typical DNA yields of 15 μg - 20 μg
  • Protocols provided for Gram + and Gram - bacteria
  • High quality genomic DNA in less than 2 hours
  • Purified DNA has an A260/A280 ratio between 1.6 and 1.9

Principle

The bacteria are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane into a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution.

The expected yield of genomic DNA will vary depending on the cell density of the bacterial culture and the bacterial species and strain used. DNA purified with the GenElute kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • W0263Wash Solution 1SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Porwollik et al.
Methods in molecular biology (Clifton, N.J.), 394, 89-103 (2008-03-28)
Microarray technology provides a convenient and relatively inexpensive way of investigating the genetic content of bacterial genomes by comparative genomic hybridization. In this method, genomic DNA of an unknown bacterial strain of interest and that of a closely related sequenced
Manuel Simões et al.
Journal of basic microbiology, 47(3), 230-242 (2007-05-24)
Biocides generally have multiple biochemical targets. Such a feature easily entangles the analysis of the mechanisms of antimicrobial action. In this study, the action of the dialdehyde biocide ortho-phtalaldehyde (OPA), on bacteria, was investigated using the Gram-negative Pseudomonas fluorescens. The
Subhajit Basu et al.
Microbial ecology, 65(4), 934-954 (2013-01-03)
A massive algal bloom of the dinoflagellate Noctiluca miliaris (green) was located in the Northern Arabian Sea by IRS-P4-2 (OCM-II) for microbiological studies, during two consecutive cruises of February-March 2009. Culturable bacterial load during bloom were ≈ 2-3-fold higher in
Hyunjin Yoon et al.
Infection and immunity, 79(1), 360-368 (2010-11-03)
Salmonella enterica serovar Typhimurium is an intracellular pathogen and a main cause of food-borne illness. In this study, a quantitative PCR (qPCR)-based competitive index (CI) method was developed to simultaneously compare the growth of multiple Salmonella strains. This method was
Joyce M Sakamoto et al.
PloS one, 9(7), e101389-e101389 (2014-07-16)
The most significant vector of tick-borne pathogens in the United States is Ixodes scapularis Say (the blacklegged tick). Previous studies have identified significant genetic, behavioral and morphological differences between northern vs. southern populations of this tick. Because tick-borne pathogens are

Articles

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Protocols

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

Related Content

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Questions

  1. Where can I find the compositions of the reagents in the bacterial genomic DNA isolation kit? Does Lysis C contain trizol?

    1 answer
    1. The formulation of the lysis solution in this kit is proprietary. However, it does not contain Trizol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service