Skip to Content
Merck
All Photos(1)

Key Documents

N8773

Sigma-Aldrich

Anti-Nerve Growth Factor-β antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-NGF-β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

capture ELISA: 0.2-0.8 μg/mL
immunohistochemistry: 5-15 μg/mL
neutralization: 0.02-0.1 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Ngf(310738)

General description

Nerve Growth factor (NGF) is a growth factor that is essential for the development and maintenance of the nervous system. NGF is composed of three non-covalently linked α, β and γ subunits. The β subunit is responsible for the biological functions of NGF. Nerve growth factor (NGF) binds to either to NGF receptor (NGFR) or the Trk receptor and can elicit biological responses through each of the receptors independently. NGF is important in the survival and differentiation of neurons and neural crest-derived cells during embryonic and early postnatal development and maintenance of peripheral neurons. NGF is also implicated in inflammation and allergic reactions . Anti-Nerve Growth Factor-β antibody recognizes rat NGF-β. The antibody shows less than 1% cross reactivity with recombinant human β-NGF.
in the presence of 10.0μg/ml of recombinat rat β-NGF using the human erythroleukemic TF-1 cell line.

Immunogen

recombinant rat β-nerve growth factor expressed in Sf 21 cells.

Application

Anti-NGF-β antibody may be used for neutralization assays; the ND50 is ~0.02-0.1 μg/ml. For immunoblotting the recommended antibody concentration is 0.1-0.2 μg/ml. The antibody is suitable for immunohistochemistry at a working concentration of 5-15 μg/ml. For ELISA, antibody concentration of 0.2-0.8 μg/ml may be used. Antibody dilution of 1:500 was used for immunoblotting of extracts of rat nerve cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uros Kovacic et al.
Neurobiology of aging, 31(2), 339-350 (2008-05-24)
Sprouting of uninjured nociceptive axons was examined in young adult, middle aged and aged rats. Axon sprouting from the spared sural nerve, both into adjacent denervated skin and into end-to-side coapted nerve graft, was significantly higher in young rats than
Bopaiah P Cheppudira et al.
BMC anesthesiology, 16(1), 73-73 (2016-09-07)
Nerve growth factor (NGF) is known to induce inflammation and pain; however its role in opioid-induced tolerance has not been studied. This study investigated the effects of an anti-NGF neutralizing antibody on the development of tolerance following chronic morphine treatment
Ana Sanchez-Rodriguez et al.
PloS one, 14(7), e0219780-e0219780 (2019-07-19)
In some induced-ovulating species, beta nerve growth factor (β-NGF) has important roles in ovulation, though data for rabbits are still inconclusive. In this study we first synthesized functional recombinant β-NGF from rabbit tissue (rrβ-NGF) to address the following objectives: 1)
Margherita Maranesi et al.
Biology of reproduction, 98(5), 634-643 (2018-02-14)
To investigate the ovulatory mechanisms triggered by raw semen (RS) in rabbits, we examined the expression of nerve growth factor (NGF)-a supposed ovulation-inducing factor (OIF)-and cognate receptors in anterior pituitary, ovary, and cervix as well as plasma NGF and luteinizing
Ana Sanchez-Rodriguez et al.
Theriogenology, 126, 206-213 (2018-12-24)
β-Nerve Growth Factor (β-NGF) is a neurotrophin which acts through its receptors TrkA and p75, performing important actions in male reproductive physiology and its presence in seminal plasma (SP) has been related to male fertility. The aim of the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service