Skip to Content
Merck
All Photos(1)

Key Documents

N8641

Sigma-Aldrich

Nutrient Mixture F-12 Ham

HEPES modification, with sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Nutrient Mixture F-12 Ham, HEPES Modification, with 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

>7.2

components

L-glutamine: no
sodium pyruvate: 0.11 g/L
NaHCO3: yes
phenol red: yes
HEPES: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Supplement with 0.146 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastasia Syntychaki et al.
Journal of proteome research, 18(9), 3521-3531 (2019-07-30)
Proteins are involved in all tasks of life, and their characterization is essential to understand the underlying mechanisms of biological processes. We present a method called "differential visual proteomics" geared to study proteome-wide structural changes of proteins and protein-complexes between
Tsuyoshi Mishiro et al.
The EMBO journal, 28(9), 1234-1245 (2009-03-27)
Long-range regulatory elements and higher-order chromatin structure coordinate the expression of multiple genes in cluster, and CTCF/cohesin-mediated chromatin insulator may be a key in this regulation. The human apolipoprotein (APO) A1/C3/A4/A5 gene region, whose alterations increase the risk of dyslipidemia
Fumitaka Kawakami et al.
PloS one, 7(1), e30834-e30834 (2012-02-04)
Leucine-rich repeat kinase 2 (LRRK2), a large protein kinase containing multi-functional domains, has been identified as the causal molecule for autosomal-dominant Parkinson's disease (PD). In the present study, we demonstrated for the first time that (i) LRRK2 interacts with tau
Henri J Huttunen et al.
Journal of molecular neuroscience : MN, 37(1), 6-15 (2008-07-12)
A common pathogenic event that occurs in all forms of Alzheimer's disease is the progressive accumulation of amyloid beta-peptide (Abeta) in brain regions responsible for higher cognitive functions. Inhibition of acyl-coenzyme A: cholesterol acyltransferase (ACAT), which generates intracellular cholesteryl esters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service