Skip to Content
Merck
All Photos(3)

Key Documents

N8509

Sigma-Aldrich

N-Nitroso-N-ethylurea

Bulk package

Synonym(s):

ENU, N-Ethyl-N-nitrosourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N3O2
CAS Number:
Molecular Weight:
117.11
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
NACRES:
NA.25

Quality Level

form

powder

contains

aqueous acetic acid as stabilizer

storage temp.

−20°C

SMILES string

CCN(N=O)C(N)=O

InChI

1S/C3H7N3O2/c1-2-6(5-8)3(4)7/h2H2,1H3,(H2,4,7)

InChI key

FUSGACRLAFQQRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Genetic determinants of blood pressure and heart rate identified through ENU-induced mutagenesis with automated meiotic mapping.: This study identifies genetic factors affecting blood pressure and heart rate using N-ethyl-N-nitrosourea (ENU)-induced mutagenesis. The research utilizes automated meiotic mapping to reveal the genetic underpinnings of these cardiovascular traits, providing insights that could inform therapeutic targets (Teixeira et al., 2024).

  • Clonality, trafficking, and molecular alterations among Hprt mutant T lymphocytes isolated from control mice versus mice treated with N-ethyl-N-nitrosourea.: This study examines the effects of ENU on T lymphocyte clonality and trafficking in mice. The findings provide insights into the mutagenic and immunological impacts of ENU, contributing to our understanding of chemical-induced mutations (Judice et al., 2023).

  • Adopting duplex sequencing technology for genetic toxicity testing: A proof-of-concept mutagenesis experiment with N-ethyl-N-nitrosourea (ENU)-exposed rats.: This proof-of-concept study integrates duplex sequencing technology to test genetic toxicity in rats exposed to ENU. The results demonstrate the feasibility and effectiveness of this approach in detecting mutagenic effects, highlighting its potential for genetic toxicity assessment (Smith-Roe et al., 2023).

Biochem/physiol Actions

DNA alkylating agent that is carcinogenic in many animal species. Induces benign and malignant tumors of numerous types, including the nervous tissue, stomach, esophagus, pancreas, respiratory tract, intestine, lymphoreticular tissues, skin, and kidney.

Packaging

Weight on dry basis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peng Huang et al.
Journal of genetics and genomics = Yi chuan xue bao, 39(9), 421-433 (2012-10-02)
Zebrafish (Danio rerio) is a well-established vertebrate animal model. A comprehensive collection of reverse genetics tools has been developed for studying gene function in this useful organism. Morpholino is the most widely used reagent to knock down target gene expression
Daniel R Beisner et al.
The Journal of experimental medicine, 210(1), 23-30 (2012-12-26)
B cell development requires tight regulation to allow for the generation of a diverse repertoire while preventing the development of autoreactive cells. We report, using N-ethyl-N-nitrosourea (ENU)-induced mutagenesis, the identification of a mutant mouse (chompB) with a block in early
Fiona C Brown et al.
Blood cells, molecules & diseases, 50(2), 86-92 (2012-10-09)
Forward genetic screens have been performed in many species to identify phenotypes in specific organ systems. We have undertaken a large-scale N-ethyl-N-nitrosourea (ENU) mutagenesis screen to identify dominant mutations that perturb erythropoiesis in mice. Mutant mice that displayed an erythrocyte
Sukalpa Chattopadhyay et al.
Journal of cell communication and signaling, 13(1), 39-52 (2018-07-07)
The evolutionarily conserved Wnt signaling pathway regulates physiological hematopoiesis, a process of formation of blood cells and has been shown to play crucial role in the development of both myeloid and lymphoid malignancies. The Wnt signaling pathway can be broadly
Ryohei Katayama et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(1), 166-174 (2014-10-30)
ROS1 rearrangement leads to constitutive ROS1 activation with potent transforming activity. In an ongoing phase I trial, the ALK tyrosine kinase inhibitor (TKI) crizotinib shows remarkable initial responses in patients with non-small cell lung cancer (NSCLC) harboring ROS1 fusions; however

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service