Skip to Content
Merck

N2250

Sigma-Aldrich

Naphthol AS-BI phosphate disodium salt hydrate

fluorogenic, ≥96.0% (HPLC), powder

Synonym(s):

2-Naphthalenecarboxamide, 7-bromo-N-(2-methoxyphenyl)-3-(phosphonooxy)-, disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13BrNNa2O6P · xH2O
CAS Number:
Molecular Weight:
496.16 (anhydrous basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Pricing and availability is not currently available.

product name

Naphthol AS-BI phosphate disodium salt hydrate, ≥96.0% (HPLC)

Quality Level

Assay

≥96.0% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to yellow

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2OP([O-])([O-])=O

InChI

1S/C18H15BrNO6P.2Na.H2O/c1-25-16-5-3-2-4-15(16)20-18(21)14-9-12-8-13(19)7-6-11(12)10-17(14)26-27(22,23)24;;;/h2-10H,1H3,(H,20,21)(H2,22,23,24);;;1H2/q;2*+1;/p-2

InChI key

WNPKMQCBZKBNOS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
N6125N2125N4006
solubility

water: 50 mg/mL, clear, colorless to yellow

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

solubility

methanol: 50 mg/mL

solubility

DMF: 50 mg/mL

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

Naphthol AS-BI (7-Bromo-3-hydroxy-2-naphtho-o-anisidine) is a substituted naphthol.[1]

Application

Naphthol AS-BI phosphate disodium salt hydrate is suitable to use:
  • in tartrate-resistant acid phosphatase (TRAP) staining to detect osteoclast activity[2]
  • as a component in Tris-HCl in staining cells to measure alkaline phosphatase (ALP) activity[3]
  • to perform alkaline phosphatase reaction[4]

Biochem/physiol Actions

Naphthol AS-BI (7-Bromo-3-hydroxy-2-naphtho-o-anisidine) can be used as a substrate to determine alkaline and acid phosphatase. It is also used to determine the activity of acid phosphatase in single cells.[1]

Substrates

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishment of human cementifying fibroma cell lines by transfection with temperature-sensitive simian virus-40 T-antigen gene and hTERT gene
Kudo Y, et al.
Bone, 30(5), 712-717 (2002)
Enhanced fibrillin-2 expression is a general feature of wound healing and sclerosis: potential alteration of cell attachment and storage of TGF-beta
Brinckmann J, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 90(5), 739-752 (2010)
Naphthol AS-BI (7-bromo-3-hydroxy-2-naphtho-o-anisidine) phosphatase and naphthol AS-BI beta-D-glucuronidase in Chinese hamster ovary cells: biochemical and flow cytometric studies.
Dolbeare FA and Phares W
The Journal of Histochemistry and Cytochemistry, 27(1), 120-124 (1979)
Effects of estrogen deficiency on microstructural changes in rat alveolar bone proper and periodontal ligament
Liu Z, et al.
Molecular Medicine Reports, 12(3), 3508-3514 (2015)
Elisheva Goldman et al.
Cells, 10(3) (2021-04-04)
Older age is associated with reduced immune function. Our aim was to study how age affects the development of apical periodontitis (AP). AP was induced in two age groups of mice (young vs. adult). Histological samples were stained by Hematoxylin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service