Skip to Content
Merck
All Photos(1)

Key Documents

N194

Sigma-Aldrich

NS-398

≥98% (HPLC), solid

Synonym(s):

N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N2O5S
CAS Number:
Molecular Weight:
314.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white

mp

127-128 °C

solubility

DMSO: >5 mg/mL
H2O: insoluble

SMILES string

CS(=O)(=O)Nc1ccc(cc1OC2CCCCC2)[N+]([O-])=O

InChI

1S/C13H18N2O5S/c1-21(18,19)14-12-8-7-10(15(16)17)9-13(12)20-11-5-3-2-4-6-11/h7-9,11,14H,2-6H2,1H3

InChI key

KTDZCOWXCWUPEO-UHFFFAOYSA-N

Gene Information

Application

NS-398 has been used as a cyclooxygenase-2 (COX2) inhibitor to study its effects on:
  • the cardiac rate in zebrafish embryos
  • apoptosis and hypoxia/reoxygenation in rat cardiomyocytes
  • the lipopolysaccharide (LPS) induced anorexia in rats

Biochem/physiol Actions

NS-398 belongs to the non-steroidal anti-inflammatory drug (NSAID) family. It exhibits anti-inflammatory, analgesic, and anti-pyretic properties.
Selective cyclooxygenase-2 (COX-2) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jihye Jung et al.
Cells, 8(7) (2019-06-30)
The epithelial-mesenchymal transition (EMT) is important in organ fibrosis. We hypothesized that growth arrest-specific protein 6 (Gas6) and its underlying mechanisms play roles in the prevention of EMT in alveolar epithelial cells (ECs). In this study, to determine whether Gas6
N Futaki et al.
General pharmacology, 24(1), 105-110 (1993-01-01)
1. NS-398 (N-[2-cyclohexyloxy-4-nitrophenyl] methanesulfonamide) is a new non-steroidal anti-inflammatory drug (NSAID) with analgesic and antipyretic effects. 2. The anti-inflammatory potency of NS-398 in rat carrageenin-induced edema was as potent as that of indomethacin and 8 times more potent than diclofenac.
Yuyin Yi et al.
Placenta, 68, 44-51 (2018-07-30)
Enhanced expression and activity of cyclooxygenase-2 (COX-2) has been reported in trophoblasts from women with preeclampsia compared with healthy pregnant women. Prostaglandin E2, the production of which is initiated by COX-2, and transforming growth factor-β1 (TGF-β1) have been shown to
Pedro Cruz Duarte et al.
Experimental neurology, 234(2), 466-481 (2012-02-09)
Brain-derived neurotrophic factor (BDNF) exists in small to medium size neurons in adult rat dorsal root ganglion (DRG) and serves as a modulator at the first synapse of the pain transmission pathway in the spinal dorsal horn. Peripheral nerve injury
Yoshihiro Morita et al.
International journal of oncology, 41(3), 885-892 (2012-06-28)
Oral squamous cell carcinoma (OSCC) is the sixth most common cancer and frequently metastasizes to the cervical lymph nodes, leading to poor survival of patients with OSCC. However, the mechanism of lymph node metastasis is not fully understood. To clarify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service