Skip to Content
Merck
All Photos(1)

Key Documents

MWGF70

Sigma-Aldrich

Gel Filtration Markers Kit for Protein Molecular Weights 6,500-66,000 Da

Synonym(s):

Gel Filtration Protein Ladder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

Quality Level

technique(s)

protein quantification: suitable

storage temp.

−20°C

General description

Gel filtration chromatography is an established method for determining the size and molecular mass of proteins. Fractionation is based on the diffusion of molecules into the pores of the resin. Larger proteins do not enter the pores of the resin as readily but pass through the fluid volume of the column faster than smaller proteins. These protein molecules elute from the column in order of decreasing molecular mass.

Application

Gel Filtration Markers Kit for Protein Molecular Weights 6,500-66,000 Da has been used as a standard in size exclusion chromatography.
Gel filtration markers are used in protein chromatography and gel filtration chromatography. Gel filtration markers have been used to study glucocorticoid-induced lymphocytolysis as a model system for apoptosis within the immune system. Gel filtration markers have also been used to isolate Polygonatum odoratum lectin, which showed remarkable anti-HSV-II activity towards Vero cells, cytotoxicity towards human melanoma A375 cells and induced apoptosis in a caspase-dependent manner.

Kit Components Also Available Separately

Product No.
Description
SDS

  • A8531Albumin, Bovine serum, 66,000 MW 1 mL/vialSDS

  • A3886Aprotinin, Bovine lung, 6,500 MW 1 mL/vialSDS

  • D4772Blue Dextran 2,000,000 MW 1 mL/vialSDS

  • C7025Carbonic Anhydrase, Bovine erythrocytes, 29,000 MW 1 mL/vialSDS

  • C7150Cytochrome c, Horse heart, 12,400 MW 1 mL/vialSDS

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filomena D'Amato et al.
The Journal of endocrinology, 197(2), 359-369 (2008-04-25)
While vg f gene knockout mice are hyperactive and hypermetabolic, surprisingly the TLQP-21 brain VGF peptide increased energy consumption, suggesting that opposing regulatory effects could be exerted by peptides alternatively cleaved from the VGF precursor. Using antisera to the VGF
N Heredia et al.
Letters in applied microbiology, 48(1), 133-139 (2008-12-06)
The mechanisms of adaptation of Clostridium perfringens to high temperatures are not well understood. In this work, the involvement of extracellular compounds in protection to heat was determined. Cells were grown in fluid thioglycollate medium or chicken broth. When mid-log
Purification and some properties of Mn peroxidase from <I>Lentinula edodes</I>.
Boer, C.G., et al.
Process. Biochem., 41(5), 1203-1207 (2006)
Miriam Kuester et al.
Journal of molecular biology, 409(2), 189-201 (2011-04-06)
Death receptors belong to the tumor necrosis factor receptor (TNFR) super family and are intimately involved in the signal transduction during apoptosis, stress response and cellular survival. Here we present the crystal structure of recombinantly expressed death receptor six (DR6)
Benjamin Blacklow et al.
Toxicon : official journal of the International Society on Toxinology, 55(6), 1171-1180 (2010-01-13)
Australo-papuan death adders (Acanthophis spp.) are a cause of serious envenomations in Papua New Guinea and northern Australia often resulting in neurotoxic paralysis. Furthermore, victims occasionally present with delayed-onset neurotoxicity that sometimes responds poorly to antivenom or anticholinesterase treatment. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service