Key Documents
MTCP7025
MTC Bio ASPIR-8 Resevoir for 8-Channel Pipettes
capacity 25 mL, non-sterile, pack of 100 ea (bulk bag)
Synonym(s):
cryo vial marker, reagent dish, reagent holder
About This Item
UNSPSC Code:
41121522
NACRES:
NB.15
sterility
non-sterile
packaging
pack of 100 ea (bulk bag)
manufacturer/tradename
MTC Bio P7025
capacity
25 mL
General description
Aspir-8 Reservoir, 25ml, 8-channel only, non-sterile, bulk pack, 100/cs
Features and Benefits
  • Graduated inner walls
  • Micro-trough U-shaped bottoms
  • Pouring spouts on all 4 corners
Legal Information
ASPIR-8 is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)