Skip to Content
Merck
All Photos(5)

Key Documents

MAK559

Sigma-Aldrich

Total Carbohydrate Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Carbohydrate Assay Kit, Carbohydrate Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104
NACRES:
NA.55

usage

sufficient for 100 colorimetric tests

Quality Level

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; metabolic disorders

storage temp.

2-8°C

General description

Carbohydrates are the most abundant biomolecules present in all living organisms. Carbohydrates perform various key roles in living organisms-fuel source (glucose), energy storage (glycogen and starch), structural component (cellulose), parts of proteins (glycoproteins, proteoglycans), components of genetic material (ribose and deoxyribose), for survival (trehalose) and more.

The Total Carbohydrate Assay Kit can be used for measuring carbohydrates in a variety of samples, including food and beverage products, solutions, cell culture supernatant and cell lines.

Total carbohydrate concentration is based on the phenol-sulfuric acid method in which polysaccharides are hydrolyzed and then converted to furfural or 5-hydroxymethyllfurfural. These compounds react with the developer to generate a chromogen, which can be detected spectrophotometrically at 490 nm.

Application

Total Carbohydrate Assay Kit has been used in the colorimetric measurement of carbohydrates in food and beverages and biological samples.

Features and Benefits

  • Simple and rapid procedure can be performed in 30 minutes
  • Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file

Suitability

Suitable for measuring total carbohydrates in adherent or suspension cells, animal and plant tissues and food products, fruit juices & other beverages.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colorimetric measurement of carbohydrates in biological wastewater treatment systems: A critical evaluation
Le C, et al.
Water Research, 94, 280-287 (2016)
Effects of monosaccharide composition on quantitative analysis of total sugar content by phenol-sulfuric acid method
Yue F, et al.
Frontiers in nutrition, 9, 963318-963318 (2022)
Estimating the carbohydrate content of various forms of tobacco by phenol-sulfuric acid method
Jain V M, et al.
Journal of education and health promotion, 6, 90-90 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service