MAK337
Aconitase Assay Kit
sufficient for 100 colorimetric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric tests
input
tissue homogenate(s)
cell lysate
serum
detection method
colorimetric
relevant disease(s)
endocrinological disorders, diabetes; neurological disorders; rheumatological diseases
storage temp.
−20°C
Related Categories
General description
Aconitase (Aconitate Hydratase) is an enzyme in the citric acid (TCA) and glyoxylate cycles that catalyzes the conversion of citrate to isocitrate. The activity of aconitase depends largely upon the iron-sulfur [Fe⁴S⁴] 2+ cluster. Related diseases include aconitase deficiency (such as myopathy and exercise intolerance. Friedreich′s ataxia and diabetes.) Mitochondrial aconitase also plays a role in stabilizing mtDNA, thereby influencing mitochondrial gene expression. Aconitase serves as a reliable marker of mitochondrial and cellular oxidative stress. The alteration of mitochondrial aconitase can lead to reduced energy production. The cytosolic form can act as an aconitase and also as an iron regulatory protein. The enzyme′s activity is inhibited by citrate analogs and fluoracetate. Oxidative stress agents like peroxynitrite, hydrogen peroxide, and superoxide can deactivate the enzyme by converting the [Fe⁴S⁴] cluster to a [Fe3S⁴] cluster.
Application
The Aconitase Assay Kit may be used for:
- Cellular and Mitchondrial Metabolism Research
- Oxidative Stress Research
- Neurological Research
Features and Benefits
- Fast and Sensitive Measurement: Achieve quantitative measurement of aconitase activity using just 20 µL of sample, with a detection range spanning from 0.5 to 100 U/L.
- High-Throughput Compatibility: Seamlessly integrate the kit into high-throughput handling systems, ensuring efficiency and accuracy in assays.
Suitability
Suitable for aconitase activity determination in biological samples (e.g. cell lysate, tissue homogenate, serum, etc.)
Principle
The Aconitase Assay Kit measures the isocitrate generated as a product of the aconitase reaction. The isocitrate is then oxidized producing NADPH and the oxidation product. The NADPH converts the dye to an intense violet color with an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to aconitase activity.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service