Skip to Content
Merck
All Photos(1)

Key Documents

M9191

Sigma-Aldrich

Microsomes from Liver, Pooled

from rat(Sprague-Dawley), female

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47

biological source

rat (Sprague-Dawley)

Quality Level

form

liquid

specific activity

≥20 units/mg protein, pH 7.4, 37 °C

packaging

vial of ~10 mg

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.

Unit Definition

One unit will reduce 1 nanomole of cytochrome C per minute at pH 7.4 at 37 deg C.
7-ethoxyresorufin O-deethylase activity: One unit will hydroxylate 1.0 picomole paclitaxel per minute at pH 7.4 at 37 deg C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service