Skip to Content
Merck

M8890

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$171.00

$171.00


Availability

Available to ship onDecember 26, 2024Details


Request a Bulk Order

Select a Size

Change View
50 MG
$171.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$171.00


Availability

Available to ship onDecember 26, 2024Details


Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

packaging

vial of 2 mL lyophilized antiserum

technique(s)

Ouchterlony double diffusion: 1:8

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
B975791466PHL89581
Betulin analytical standard

92648

Betulin

Betulin ≥98%

B9757

Betulin

Betulin phyproof® Reference Substance

PHL89581

Betulin

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

format

neat

format

-

format

neat

format

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

grade

analytical standard

grade

-

grade

analytical standard

grade

primary reference standard

assay

≥97.5% (TLC)

assay

≥98%

assay

≥97.0% (HPLC)

assay

≥95.0% (HPLC)

Application

Anti-Mouse IgG (whole molecule) antibody produced in goat was used to immunoprecipitate bovine brain adenylyl cyclase. It was used α-defensin isoforms in the plasma of type I diabetic patients by radio immune assay.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 mL deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julius Grzeschik et al.
Biotechnology journal, 14(4), e1800466-e1800466 (2018-10-24)
Yeast surface display emerged as a viable tool for the generation of human and murine monoclonal antibodies. This platform technology enables the careful definition of selection conditions, the potential for high-throughput screening, as well as the isolation of antibodies recognizing
Adrian Elter et al.
Scientific reports, 11(1), 7880-7880 (2021-04-14)
Since the pandemic outbreak of Covid-19 in December 2019, several lateral flow assay (LFA) devices were developed to enable the constant monitoring of regional and global infection processes. Additionally, innumerable lateral flow test devices are frequently used for determination of
S Mollner et al.
European journal of biochemistry, 171(1-2), 265-271 (1988-01-15)
Four monoclonal antibodies, raised against the 115-kDa adenylyl cyclase from bovine brain [Pfeuffer, E. et al. (1985) EMBO J. 4, 3675-3679] have been selected and designated BBC-1 to BBC-4. BBC-1 and BBC-3 are highly specific for the 115-kDa enzyme from
M Saraheimo et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 23(3), 914-918 (2007-11-16)
Diabetic nephropathy is associated with low-grade inflammation and activation of the complement system. Defensins, as part of the innate immune system, may play a regulatory role in the complement cascade and may also augment the production of proinflammatory cytokines. The
S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service