Skip to Content
Merck
All Photos(1)

Key Documents

M7927

Sigma-Aldrich

Anti-MAP Kinase (ERK-1, 351-368) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen, ERK-1 44 kDa
antigen, ERK-2 42 kDa

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): 5 μg using NIH 3T3 mouse fibroblast lysates
microarray: suitable
western blot: 1:5,000 using NIH 3T3 mouse fibroblast lysate or rat brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK3(5595)
mouse ... Mapk3(26417)
rat ... Mapk3(50689)

General description

Mitogen-activated protein kinase 3 (ERK1) is mapped to human chromosome 16p11.2. It has a serine/threonine domain and a transactivation domain (TAD) and a nuclear localization sequence (NLS).

Specificity

By immunoblotting, the antibody reacts with ERK-1 and ERK-2.

Immunogen

synthetic peptide corresponding to amino acids 351-368 of human ERK-1.

Application

Anti-MAP Kinase (ERK-1, 351-368) antibody produced in rabbit has been used in:
  • immunofluorescence
  • immunoprecipitation
  • western blotting.

Biochem/physiol Actions

Mitogen-activated protein kinase (MAPK) superfamily of enzymes is involved in widespread signalling pathways. Members of this family include the ERK1/2 (extracellular signal-regulated protein kinase, also termed p42/p44 MAPK), c-Jun N-terminal kinases (JNK) and p38 MAPK subfamilies. These are the terminal enzymes in a signalling cascade where each kinase phosphorylates and activates the next member in the sequence. Phosphorylation of both tyrosine and threonine is essential for the full activation of all MAPKs. Several kinases participate in activation of the ERK cascade. This cascade is initiated by the small G protein Ras, which upon stimulation causes activation Raf1 kinase. Raf1 continues the transmission by activating MEK. Activated MEK appears to be the only kinase capable of specifically phosphorylating and activating ERK. ERK1 (p44) and ERK2 (p42) require the dual phosphorylation in the catalytic kinase domain by MEKs for their full activity. ERK1 is phosphorylated on Tyr204 and Thr202, and ERK2 on Tyr187 and Thr185. ERK1 and 2 may also undergo autophosphorylation on these residues. ERK appears to be an important regulatory molecule, which by can phosphorylate regulatory targets in the cytosol (phospholipase A2, PLA2), translocated into and phosphorylate substrates in the nucleus (ELK1). The activation of ERK cascade mediates and regulates the signal transduction pathways in response to stress, mitogenic signals and is important in development and differentiation, learning, memory and survival.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p42/p44 MAPKs are intracellular targets of the CDK inhibitor purvalanol
Knockaert M, et al.
Oncogene, 21(42), 6413-6413 (2002)
Frequency modulation of ERK activation dynamics rewires cell fate
Ryu H, et al.
Molecular Systems Biology, 11(11), 50-83 (2015)
Association and mutation analyses of 16p11. 2 autism candidate genes
Kumar RA, et al.
Testing, 4(2), e4582-e4582 (2009)
Prevention of glucocorticoid-induced apoptosis in osteocytes and osteoblasts by calbindin-D28k
Liu Y, et al.
Journal of Bone and Mineral Research, 19(3), 479-490 (2004)
Yan Liu et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 19(3), 479-490 (2004-03-26)
This study show for the first time that calbindin-D28k can prevent glucocorticoid-induced bone cell death. The anti-apoptotic effect of calbindin-D28k involves inhibition of glucocorticoid induced caspase 3 activation as well as ERK activation. Recent studies have indicated that deleterious effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service