Skip to Content
Merck
All Photos(1)

Key Documents

M7894

Sigma-Aldrich

IgG1, Kappa from murine myeloma

clone MOPC 21, ascites fluid, lyophilized powder

Synonym(s):

Mouse IgG1-κ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

clone

MOPC 21, monoclonal

form

lyophilized powder

application(s)

research pathology

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

The MOPC-21 tumor line that produces mouse IgG1, κ is a mineral oil induced plasmacytoma originated and carried intraperitoneally in BALB/c mice. Immunoglobulins G are made up of two identical light chains and two identical heavy chains, that are covalently connected by interchain disulphide bonds. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b, IgG2c and IgG3.

Application

IgG1, Kappa from murine myeloma has been used:
  • in stimulation assay and enzyme-linked immunosorbent assay (ELISA)
  • as isotype controls in islet hormone production
  • in flow cytometric analysis, in cell migration assay
  • as control antibody in evaluation of methylation

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. It provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Reconstitution

Reconstitute with 1 mL deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-6 promotes malignant growth of skin SCCs by regulating a network of autocrine and paracrine cytokines
Lederle W, et al.
International Journal of Cancer. Journal International Du Cancer, 128(12), 2803-2814 (2011)
J Zhu et al.
American journal of respiratory and critical care medicine, 164(12), 2220-2228 (2001-12-26)
We wished to determine if the inflammatory cells surrounding the airway mucus-secreting glands in chronic bronchitis (CB) were associated with interleukin (IL)-4 and IL-5 mRNA expression and whether the CD8 T cell population expressed these cytokines. Digoxigenin-labeled IL-4 and IL-5
M-ficolin concentrations in cord blood are related to circulating phagocytes and to early-onset sepsis
Schlapbach LJ, et al.
Pediatric Research, 71(4-1), 368-368 (2012)
Eva Obermueller et al.
Cancer research, 64(21), 7801-7812 (2004-11-03)
Tumor growth and progression are critically controlled by alterations in the microenvironment often caused by an aberrant expression of growth factors and receptors. We demonstrated previously that tumor progression in patients and in the experimental HaCaT tumor model for skin
The Laboratory Rat (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service