Skip to Content
Merck
All Photos(4)

Key Documents

M6020

Sigma-Aldrich

D-(+)-Mannose

powder, BioReagent, suitable for cell culture

Synonym(s):

D-Mannopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1564373
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

biological source

plant (Spruce, Birch or Beech Wood, Glucose)

Quality Level

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

133-140 °C (lit.)

solubility

H2O: 50 mg/mL

application(s)

agriculture

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mannose is an aldohexose carbohydrate. It is isomer of glucose with varying C2 position configuration. Majority of mannose is synthesised from its epimer glucose. It is part of the glycans present in endoplasmic reticulum (ER)/Golgi.

Application

D-(+)-Mannose has been used:
  • as a reference standard in monosaccharide analysis and quantification from sea squirts Ascidiella aspersa
  • as a medium component for the selection of P898012 sorgum seeds transfected with Agrobacterium
  • in segmented filamentous bacterium (SFB) medium supplement for culturing human cell lines

Mannose is an aldohexose carbohydrate. This product may be used to study the effects of exogenous mannose on in vitro processes such as glycosylation.

Biochem/physiol Actions

Mannose may be used to study the effects of exogenous mannose on in vitro processes such as glycosylation. cis-Golgi glycoproteins have high mannose and are prone to Endoglycosidase H or peptide N-glycosidase (N-glycanase) digestion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic manipulation of glycosylation disorders in humans and animal models
Freeze HH and Sharma V
Seminars in Cell & Developmental Biology, 21(6), 655-662 (2010)
Agrobacterium-mediated transformation of Sorghum bicolor using immature embryos
Transgenic Plants, 109-122 (2012)
Essentials of Carbohydrate Chemistry by John F. Robyt
Pendarvis R
The Chemical Educator, 3(5), 1-2 (1998)
M J Carlino et al.
Gut microbes, 12(1), 1-21 (2020-11-03)
Infectious diarrhea causes approximately 179 million illnesses annually in the US. Multiplex PCR assays for enteric pathogens detect enteropathogenic Escherichia coli (EPEC) in 12-29% of diarrheal stool samples from all age groups in developed nations. The aim of this study
Growth and host interaction of mouse segmented filamentous bacteria in vitro
Schnupf P, et al.
Nature, 520(7545), 99-100 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service