Skip to Content
Merck
All Photos(1)

Key Documents

M5899

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) antibody produced in goat

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:70,000
quantitative precipitin assay: 3.2-4.8 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.

Specificity

The antiserum is determined to be immunospecific for mouse IgG by immunoelectrophoresis (IEP) against normal mouse serum and purified mouse IgG.

Application

Anti-Mouse IgG (whole molecule) antibody produced in goat has been used as secondary antibody in:
  • immunoblotting
  • immunohistochemistry
  • proteomic microarrays
Anti-Mouse IgG (whole molecule) antibody produced in goat has been used in:
  • the preparation of immunochromatography strips for magneto-enzyme lateral flow immunoassay
  • immunochromatography
  • MYCN DNA-binding analysis (ChIP-seq)

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

IgG antibody provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Immunoglobulin G (IgG) helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Quality

The antiserum has been treated to remove lipoproteins.

Physical form

Supplied as a liquid containing 15 mM sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tien V Tran et al.
PeerJ, 7, e7779-e7779 (2019-10-04)
Dengue infection represents a global health issue of growing importance. Dengue non-structural protein 1 (NS1) plays a central role in the early detection of the disease. The most common method for NS1 detection is testing by lateral flow immunoassays (LFIAs)
David J Duffy et al.
Oncotarget, 7(37), 60310-60331 (2016-08-18)
Wnt signalling is involved in the formation, metastasis and relapse of a wide array of cancers. However, there is ongoing debate as to whether activation or inhibition of the pathway holds the most promise as a therapeutic treatment for cancer
Katja C Erlach et al.
Journal of virology, 76(6), 2857-2870 (2002-02-28)
Tumor relapse and cytomegalovirus (CMV) infection are major concerns in the therapy of hematopoietic malignancies by bone marrow transplantation (BMT). Little attention so far has been given to a possible pathogenetic interplay between CMV and lymphomas. CMV inhibits stem cell
Barbara Wróblewska et al.
Nutrients, 14(20) (2022-10-28)
Barley (Hordeum vulgare L.) proteins are taxonomically homologous to wheat proteins and react with sera from patients with baker's asthma. In the current work, the crude extract of barley proteins was divided into six fractions on DEAE-Sepharose. Their immunoreactivity in
Yi Xu et al.
Oncology letters, 15(6), 9117-9125 (2018-05-29)
Chrysin is a natural flavone that has various biological activities, including antitumor effects. However, the effect of chrysin on bladder cancer cells remains elusive. The present study investigated the effects of chrysin on bladder cancer cells and its underlying mechanisms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service