Skip to Content
Merck
All Photos(1)

Key Documents

M5449

Sigma-Aldrich

MKT-077

≥98% (HPLC), powder

Synonym(s):

1-Ethyl-2-[[3-ethyl-5-(3-methyl-2(3H)-benzothiazolylidene)-4-oxo-2-thiazolidinylidene]methyl]-pyridinium chloride, FJ-776

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H22N3OS2Cl
CAS Number:
Molecular Weight:
432.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

orange to red

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=C(S/2)C=CC=C1)C2=C3C(N(CC)/C(S\3)=C/C4=[N+](CC)C=CC=C4)=O.[Cl-]

InChI

1S/C21H22N3OS2.ClH/c1-4-23-13-9-8-10-15(23)14-18-24(5-2)20(25)19(27-18)21-22(3)16-11-6-7-12-17(16)26-21;/h6-14H,4-5H2,1-3H3;1H/q+1;/p-1/b21-19+;

InChI key

VSKYOTRJSLYFHX-UXJRWBAGSA-M

Biochem/physiol Actions

MKT-077 is an inhibitor of mitochondrial hsp70 family member, mortalin (mot-2)n; causes selective death of cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Edan Dhamad et al.
American journal of physiology. Cell physiology, 318(2), C289-C303 (2019-12-05)
Glucose-regulated protein 75 (GRP75) was first characterized in mammals as a heat shock protein-70 (HSP70) family stress chaperone based on its sequence homology. Extensive studies in mammals showed that GRP75 is induced by various stressors such as glucose deprivation, oxidative
Sunil Joseph et al.
Biology open, 6(4), 481-488 (2017-03-16)
Earlier studies suggested that
Sujit Pujhari et al.
Emerging microbes & infections, 8(1), 8-16 (2019-03-15)
Zika virus (ZIKV) is a historically neglected mosquito-borne flavivirus that has caused recent epidemics in the western hemisphere. ZIKV has been associated with severe symptoms including infant microcephaly and Guillain-Barré syndrome, stimulating interest in understanding factors governing ZIKV infection. Heat
Jemma Day et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 14611-14624 (2019-11-07)
Plasmodium falciparum is the most lethal of human-infective malaria parasites. A hallmark of P. falciparum malaria is extensive remodeling of host erythrocytes by the parasite, which facilitates the development of virulence properties such as host cell adhesion to the endothelial
Tuoen Liu et al.
PloS one, 12(2), e0170470-e0170470 (2017-02-09)
Myelodysplastic syndromes (MDS) are the most common adult myeloid blood cancers in the US. Patients have increased apoptosis in their bone marrow cells leading to low peripheral blood counts. The full complement of gene mutations that contribute to increased apoptosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service