M4278
Monoclonal Anti-MAP1 antibody produced in mouse
clone HM-1, ascites fluid
Synonym(s):
Anti-MAP1a
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
HM-1, monoclonal
contains
15 mM sodium azide
species reactivity
mouse, rat
technique(s)
immunohistochemistry (frozen sections): suitable
microarray: suitable
western blot: 1:500 using a fresh total rat brain extract or an enriched microtubule protein preparation
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Mtap1a(17754)
rat ... Map1a(25152)
Related Categories
General description
Specificity
Immunogen
Application
- immunoblotting
- dot blot
- immunocytochemistry
- immunohistochemistry
Biochem/physiol Actions
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service