Skip to Content
Merck
All Photos(1)

Key Documents

M3696

Sigma-Aldrich

Anti-MAP2 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Microtubule associated protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~280 kDa
antigen ~72 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

flow cytometry: 1:200-1:500
immunohistochemistry: 1:100-1:200
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP2(4133)
mouse ... Mtap2(17756)
rat ... Map2(25595)

Related Categories

General description

MAP2 is a microtubule-associated protein that regulates the structure and function of the cytoskeleton. The protein also modulates morphogenesis of neurons and cell proliferation. Interactions of MAP2 with F-actin have been implicated in neurite initiation and microtubular re-organization. Decreased levels of MAP2 have been associated with transmissible spongiform encephalopathies . Anti-MAP2 antibody is specific for MAP2 in human, mice and rats.
Microtubule associated protein 2 (MAP2) is a neuron specific protein, which is highly expressed in the central nervous system. It is the member of cytoskeletal proteins. The gene is located on human chromosome 2q34.

Immunogen

synthetic peptide corresponding to residues 2-15 of human MAP2.

Application

Anti-MAP2 antibody is suitable for use in immunohistochemistry (1:500 using Zamboni′s fixative-fixed, frozen mouse spinal cord and thalamus sections) , flow cytometry (1:200-1:500), immunoblotting (approx. 72 kDa and 280 kDa) and western blot (1:500-1:1,000).
Anti-MAP2 antibody produced in rabbit has been used in immunocytochemistry.

Biochem/physiol Actions

Microtubule associated protein 2 (MAP2) plays a critical role in the formation of dendritic protrusions in astrocytes and oligodendrocytes. It maintains the homeostasis of microtubules. The protein plays an important role in neuronal plasticity.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional upregulation of microtubule-associated protein 2 is involved in the protein kinase A-induced decrease in the invasiveness of glioma cells
Zhou Y, et al.
Neuro-Oncology, 17(12), 1578-1588 (2015)
Cory C Toth et al.
Molecular pain, 6, 16-16 (2010-03-20)
Despite the frequency of diabetes mellitus and its relationship to diabetic peripheral neuropathy (DPN) and neuropathic pain (NeP), our understanding of underlying mechanisms leading to chronic pain in diabetes remains poor. Recent evidence has demonstated a prominent role of microglial
Specific binding of dehydroepiandrosterone to the N terminus of the microtubule-associated protein MAP2
Laurine E, et al.
The Journal of Biological Chemistry, 278(32), 29979-29986 (2003)
Rouhollah Habibey et al.
Scientific reports, 7(1), 8558-8558 (2017-08-19)
Due to their small dimensions, electrophysiology on thin and intricate axonal branches in support of understanding their role in normal and diseased brain function poses experimental challenges. To reduce experimental complexity, we coupled microelectrode arrays (MEAs) to bi-level microchannel devices
Qinghai Liu et al.
PloS one, 11(4), e0153376-e0153376 (2016-04-14)
This study examines the effects of nano-size particulate matter (nPM) exposure in the setting of murine reperfused stroke. Particulate matter is a potent source of inflammation and oxidative stress. These processes are known to influence stroke progression through recruitment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service