Skip to Content
Merck
All Photos(1)

Key Documents

M265

Sigma-Aldrich

Anti-Glutamate Receptor NMDAR2B (NR2B) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-DEE27, Anti-EIEE27, Anti-GluN2B, Anti-MRD6, Anti-NMDAR2B, Anti-NR2B, Anti-NR3, Anti-hNR3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen 180 kDa

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry (frozen sections): 1:1,000
immunoprecipitation (IP): 3 μL using 200 μg of rat brain
western blot: 1 μg/mL using 35S-Protein A (with ECL dilutions may be higher)

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIN2B(2904)
mouse ... Grin2b(14812)
rat ... Grin2b(24410)

General description

Glutamate ionotropic receptor N-methyl-D-aspartate (NMDA) type subunit 2B (GRIN2B) codes for N-methyl D-aspartate receptor subtype 2B (GluN2B). It is an important constituent of the N-methyl D-aspartate receptor protein. GRIN2B gene is located on human chromosome 12p13.1.

Immunogen

C-terminal portion of NR2B (amino acids 984-1104).

Application

Anti-Glutamate Receptor NMDAR2B (NR2B) antibody is suitable for use in immunohistochemistry (1:1,000 dilution, using frozen sections), immunoprecipitation (3 μL using 200 μg of rat brain), and indirect ELISA. The antibody may also be used for western blot (1 μg/mL using (35S)-Protein A) and immunoblot (180 kDa).
Anti-Glutamate Receptor NMDAR2B (NR2B) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Glutamate ionotropic receptor N-methyl-D-aspartate (NMDA) type subunit 2B (GRIN2B) plays a key role in synaptic plasticity, language acquisition, neural pattern formation and short-term memory. Mutations on GRIN2B gene results in Parkinson′s disease, schizophrenia, bipolar disorder, Alzheimer′s disease and obsessive-compulsive disorder.
The functional N-Methyl-D-Aspartate (NMDA) receptor complex consists of a NMDAR1 (NR1) subunit and atleast one NMDAR2 subunit ranging from NR2A-NR2D. These receptor complexes have been implicated in cochlear functions and neuronal plasticity. The C-terminal end of NR2B receptor may regulate synaptic localization.

Physical form

Lyophilized from ammonium bicarbonate (5 mM).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steve Standley et al.
PloS one, 7(6), e39585-e39585 (2012-07-05)
NMDA receptor NR2A/B subunits have PDZ-binding domains on their extreme C-termini that are known to interact with the PSD-95 family and other PDZ proteins. We explore the interactions between PSD-95 family proteins and the NR2A/B cytoplasmic tails, and the consequences
D Joelson et al.
Microscopy research and technique, 41(3), 246-262 (1998-05-30)
The distribution of immunoreactivity for the ionotropic N-methyl-D-aspartate (NMDA) receptor subunits was mapped in the cochlear nucleus of postnatal day (P) 7, P14, P21, and P28 gerbils. Frozen sections and serial plastic sections of tissue were incubated with antibodies to
Mi Zhou et al.
Neuroscience, 376, 24-39 (2018-02-17)
The amygdala plays a key role in the pathophysiology of depression, but the molecular mechanisms underlying amygdalar hyperactivity in depression remain unclear. In this study, we used a chronic mild stress (CMS) protocol to separate susceptible and insusceptible rat subgroups.
Maowen Ba et al.
Behavioural brain research, 356, 41-45 (2018-08-22)
Augmented function of N-methyl-d-aspartate receptor subunit 2B (NR2B) and Src protein tyrosine kinase have been demonstrated to get involved in the pathological mechanisms of dyskinesia. In view of functional interactions between NR2B and Src, we investigated the effects of uncoupling
Naoya Morisada et al.
Human genome variation, 3, 16029-16029 (2016-09-23)
N-methyl D-aspartate receptor subtype 2B (GluN2B), encoded by GRIN2B, is one of the components of the N-methyl D-aspartate receptor protein. Aberrations in GRIN2B have been reported to be responsible for various types of neurodevelopmental disorders. We report a Japanese boy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service