Skip to Content
Merck
All Photos(1)

Key Documents

M2195

Sigma-Aldrich

MOCPAC

≥95% (HPLC), solid

Synonym(s):

Benzyl (S)-[1-(4-methyl-2-oxo-2H-chromen-7-ylcarbamoyl)-5-propionylaminopentyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N3O6
CAS Number:
Molecular Weight:
493.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

solubility

DMSO: soluble 21 mg/mL

storage temp.

−20°C

SMILES string

CCC(=O)NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C27H31N3O6/c1-3-24(31)28-14-8-7-11-22(30-27(34)35-17-19-9-5-4-6-10-19)26(33)29-20-12-13-21-18(2)15-25(32)36-23(21)16-20/h4-6,9-10,12-13,15-16,22H,3,7-8,11,14,17H2,1-2H3,(H,28,31)(H,29,33)(H,30,34)/t22-/m0/s1

InChI key

BFDGUJKFQRJHJM-QFIPXVFZSA-N

Biochem/physiol Actions

Selective HDAC1 substrate (over HDAC6, class I over class II).

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service