Skip to Content
Merck
All Photos(1)

Key Documents

M1796

Sigma-Aldrich

(±)-3,4-Methylenedioxy-N-­ethyl­amphetamine hydrochloride

≥98% (TLC)

Synonym(s):

MDEA hydrochloride, N-Ethyl-3,4-MDA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO2 · HCl
CAS Number:
Molecular Weight:
243.73
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

SMILES string

Cl[H].NCCc1ccc2OCOc2c1

InChI

1S/C12H17NO2.ClH/c1-3-13-9(2)6-10-4-5-11-12(7-10)15-8-14-11;/h4-5,7,9,13H,3,6,8H2,1-2H3;1H

InChI key

IBDIPBWIXJRJQM-UHFFFAOYSA-N

Biochem/physiol Actions

Psychotropic compound causing a reduction in serotonin level and in tryptophan hydroxylase activity in the brain. May cause structural damage to serotonenergic neurons.

Other Notes

CAS# 116861-63-3, which was listed previously, is for histidine-copper complex.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Johnson et al.
Biochemical pharmacology, 38(23), 4333-4338 (1989-12-01)
The effect of N-ethyl-3,4-methylenedioxyamphetamine (MDE) on the central serotonergic system was studied. Within 1 hr after administration of MDE (10 mg/kg), the concentration of 5-hydroxytryptamine (5-HT) and the activity of tryptophan hydroxylase (TPH) had declined significantly in the hippocampus but
H G Series et al.
Experimental neurology, 128(1), 50-58 (1994-07-01)
N-ethyl-3,4-methylenedioxyamphetamine (MDE) is one of a group of substituted amphetamines which have effects on several serotonergic markers such as tissue levels of serotonin and activity of tryptophan hydroxylase. In this study we have compared its effects on the rat brain
Tsadik T Abraham et al.
Journal of analytical toxicology, 33(8), 439-446 (2009-10-31)
3,4-Methylenedioxymethamphetamine (MDMA), or ecstasy, is excreted as unchanged drug, 3,4-methylenedioxyamphetamine (MDA), and free and glucuronidated/sulfated 4-hydroxy-3-methoxymethamphetamine (HMMA), and 4-hydroxy-3-methoxyamphetamine (HMA) metabolites. The aim of this paper is to describe the pattern and timeframe of excretion of MDMA and its metabolites
Markus R Meyer et al.
Drug metabolism and disposition: the biological fate of chemicals, 37(6), 1152-1156 (2009-03-21)
The 3,4-methylenedioxy-methamphetamine (MDMA)-related designer drug 3,4-methylenedioxyethylamphetamine (MDEA, Eve) is a chiral compound that is mainly metabolized by N-deethylation and demethylenation during phase I metabolism. The involvement of several cytochrome P450 (P450) isozymes in these metabolic steps has been demonstrated by
Peter R Stout et al.
Journal of analytical toxicology, 34(8), 430-443 (2010-01-01)
On November 25, 2008, the U.S. Department of Health and Human Services posted a final notice in the Federal Register authorizing the use of liquid chromatography-tandem mass spectrometry (LC-MS-MS) and other technologies in federally regulated workplace drug testing (WPDT) programs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service