M1686
Greiner 384 well plates, polypropylene
120 μL/well, flat bottom natural polypropylene wells
Synonym(s):
384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates
About This Item
flat bottom natural polypropylene wells
polypropylene
Recommended Products
material
clear polypropylene plate
flat bottom natural polypropylene wells
polypropylene
sterility
non-sterile
feature
lid: no
packaging
case of 100
manufacturer/tradename
Greiner 781201
maximum volume
152 μL
size
384 wells
working volume
145 μL
color
natural
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Appropriate for applications requiring increased chemical resistance or heat stability, e.g. liquid storage or compound libraries in drug screening.
Other Notes
Suitability
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service