Skip to Content
Merck
All Photos(1)

Key Documents

M1574

Sigma-Aldrich

Miglitol

Synonym(s):

1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO5
CAS Number:
Molecular Weight:
207.22
EC Number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

mp

142-147  °C

solubility

water: 5 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

N1([C@@H]([C@H]([C@@H]([C@H](C1)O)O)O)CO)CCO

InChI

1S/C8H17NO5/c10-2-1-9-3-6(12)8(14)7(13)5(9)4-11/h5-8,10-14H,1-4H2/t5-,6+,7-,8-/m1/s1

InChI key

IBAQFPQHRJAVAV-ULAWRXDQSA-N

General description

Miglitol is an alpha-glucosidase inhibitor used as a glucose-lowering drug in diabetes research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuki Mochizuki et al.
Bioscience, biotechnology, and biochemistry, 74(12), 2470-2474 (2010-12-15)
β(2) integrins (CD11s/CD18) promote the attachment of leukocytes to vascular endothelial cells. We performed in this study sucrose loading to rats with moderate postprandial hyperglycemia with/without once-daily dosing of the α-glucosidase inhibitor, miglitol, for 4 days under 4-h fasting conditions.
Kazuki Mochizuki et al.
Metabolism: clinical and experimental, 60(11), 1560-1565 (2011-05-10)
Otsuka Long-Evans Tokushima Fatty (OLETF) rats, an animal model of type 2 diabetes mellitus, exhibit chronic and slowly progressive hyperglycemia with obesity. In this study, we examined whether dietary supplementation with the α-glucosidase inhibitor miglitol from the preonset stage improves
Kyra Jones et al.
Bioorganic & medicinal chemistry, 19(13), 3929-3934 (2011-06-15)
Inhibition of intestinal α-glucosidases and pancreatic α-amylases is an approach to controlling blood glucose and serum insulin levels in individuals with Type II diabetes. The two human intestinal glucosidases are maltase-glucoamylase and sucrase-isomaltase. Each incorporates two family 31 glycoside hydrolases
Kenichi Matsuura et al.
Diabetes technology & therapeutics, 14(5), 423-429 (2012-02-10)
We compared the effects of miglitol as an add-on to bolus insulin and dose-intensified bolus insulin on postprandial glycemic excursions by continuous glucose monitoring (CGM). The glucose levels of 21 type 2 diabetes patients admitted for glycemic control were monitored
Takuo Emoto et al.
The American journal of cardiology, 109(1), 42-46 (2011-09-29)
Repeated postprandial hyperglycemia may play an important role in the development of atherosclerosis by suppressing vascular endothelial function. Although miglitol suppresses the elevation of blood glucose levels shortly after a meal more than other α-glucosidase inhibitors, the effect of 3-month

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service