Skip to Content
Merck
All Photos(1)

Key Documents

M0418

Sigma-Aldrich

Melanin

BioReagent, Synthetic, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic

product line

BioReagent

Assay

≥97% (TLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

1 N NH4OH: 20 mg/mL

storage temp.

−20°C

SMILES string

[nH]1c2c3c(c4c5c([nH]c4)c([c]([c](c5c3c1)=O)=O)C)[c]([c](c2C)=O)=O

InChI

1S/C18H10N2O4/c1-5-13-9-7(3-19-13)12-10-8(11(9)17(23)15(5)21)4-20-14(10)6(2)16(22)18(12)24/h3-4,19-20H,1-2H3

InChI key

XUMBMVFBXHLACL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Melanin is a distinctive class of pigment found in animals, plants, bacteria, and fungi. It is an amorphous polymer formed by the polymerization of phenolic and indolic compounds.

Application

Melanin has been used:
  • to incubate normal human keratinocytes for Masson-Fontana staining
  • as a standard in the determination of melanin content in normal human epidermal melanocytes and immortalized human keratinocytes
  • as an absorber to mimic human skin or melanocytic nevi and melanoma

Biochem/physiol Actions

Melanin plays a role in camouflage and energy harvesting. It also helps in protection against ionizing UV radiations, protection against biotic factors such as host defense against pathogens, and abiotic factors like heat, cold and osmotic stress. Melanin acts as a potent cation chelator and is a component of several photoprotective creams as it facilitates free radical scavenging activity. Melanin may be useful as a photo protectant in cell culture.

Preparation Note

Prepared by oxidation of tyrosine with hydrogen peroxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadine Santana-Magal et al.
Cancer research, 80(10), 1942-1956 (2020-03-05)
The recent success of checkpoint blockade therapies has established immunotherapy as one of the most promising treatments for melanoma. Nonetheless, a complete curative response following immunotherapy is observed only in a fraction of patients. To identify what factors limit the
Michael Waisberg et al.
PloS one, 7(1), e29493-e29493 (2012-01-14)
Malaria has had the largest impact of any infectious disease on shaping the human genome, exerting enormous selective pressure on genes that improve survival in severe malaria infections. Modern humans originated in Africa and lost skin melanization as they migrated
Naira V Margaryan et al.
International journal of molecular sciences, 17(3), 418-418 (2016-03-25)
Expression of Nodal, a Transforming Growth Factor-beta (TGF-β) related growth factor, is associated with aggressive melanoma. Nodal expression in adult dysplastic nevi may predict the development of aggressive melanoma in some patients. A subset of pediatric patients diagnosed with giant
Li Ni-Komatsu et al.
Molecular pharmacology, 74(6), 1576-1586 (2008-09-20)
A series of quinolines, including chloroquine and quinine, were identified as potent pigmentation inhibitors through screening a compound library in murine melanocytes. Structure-activity relationship analysis indicated that 4-substituted amino groups with a tertiary amine side chain, such as chloroquine, were

Questions

  1. I wonder if you can let me know what type of melanin pigment this reagent has. Is it pheo-, eu- or mixed melanin? What is the level of polymerization? Thank you!

    1 answer
    1. This product is synthetic melanin, per Med. Res. Reviews 8 (1988) pp. 525-556, synthetic melanins are "eumelanin type pigments". Unfortunately, no additional information on the level of polymerization is available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service