Skip to Content
Merck
All Photos(1)

Key Documents

L9755

Sigma-Aldrich

Sodium dodecanoate

99-100%

Synonym(s):

Dodecanoic acid sodium salt, Lauric acid sodium salt, Sodium laurate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COONa
CAS Number:
Molecular Weight:
222.30
Beilstein:
3574124
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

99-100%

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].CCCCCCCCCCCC([O-])=O

InChI

1S/C12H24O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12(13)14;/h2-11H2,1H3,(H,13,14);/q;+1/p-1

InChI key

BTURAGWYSMTVOW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium dodecanoate (sodium laurate) is used in cholloidal chemistry as a nonionic surfactant.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenzo Lisuzzo et al.
Journal of colloid and interface science, 547, 361-369 (2019-04-12)
The filling of halloysite nanotubes with active compounds solubilized in aqueous solvent was investigated theoretically and experimentally. Based on Knudsen thermogravimetric data, we demonstrated the water confinement within the cavity of halloysite. This process is crucial to properly describe the
Jon G Hansted et al.
Biochimica et biophysica acta, 1814(5), 713-723 (2011-03-29)
The milk protein β-lactoglobulin (βLG) dominates the properties of whey aggregates in food products. Here we use spectroscopic and calorimetric techniques to elucidate how anionic, cationic and non-ionic surfactants interact with bovine βLG and modulate its heat-induced aggregation. Alkyl trimethyl
Isabel Dittmann et al.
Pharmaceutical research, 31(9), 2539-2548 (2014-03-19)
To mechanistically analyze effects of the medium-chain fatty acid laurate on transepithelial permeability in confluent monolayers of the intestinal epithelial cell line HT-29/B6, in context with an application as an absorption enhancer improving transepithelial drug permeation. Transepithelial resistance and apparent
David J K Swainsbury et al.
Science advances, 7(3) (2021-02-02)
The reaction-center light-harvesting complex 1 (RC-LH1) is the core photosynthetic component in purple phototrophic bacteria. We present two cryo-electron microscopy structures of RC-LH1 complexes from Rhodopseudomonas palustris A 2.65-Å resolution structure of the RC-LH114-W complex consists of an open 14-subunit
Cs Kotsmar et al.
Advances in colloid and interface science, 150(1), 41-54 (2009-06-06)
Depending on the bulk composition, adsorption layers formed from mixed protein/surfactant solutions contain different amounts of protein. Clearly, increasing amounts of surfactant should decrease the amount of adsorbed proteins successively. However, due to the much larger adsorption energy, proteins are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service