Skip to Content
Merck
All Photos(1)

Key Documents

L5139

Sigma-Aldrich

Lycorine hydrochloride

powder, ≥98% (TLC)

Synonym(s):

Licorin hydrochloride, Lychorine chloride, Lycorine HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO4 · HCl
CAS Number:
Molecular Weight:
323.77
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Lycorine hydrochloride, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

mp

210-212  °C

solubility

ethanol: 10 mg/mL, clear, colorless to very faintly yellow

storage temp.

2-8°C

SMILES string

Cl.O[C@H]1C=C2CCN3Cc4cc5OCOc5cc4[C@H]([C@@H]1O)[C@@H]23

InChI

1S/C16H17NO4.ClH/c18-11-3-8-1-2-17-6-9-4-12-13(21-7-20-12)5-10(9)14(15(8)17)16(11)19;/h3-5,11,14-16,18-19H,1-2,6-7H2;1H/t11-,14-,15+,16+;/m0./s1

InChI key

VUVNTYCHKZBOMV-NVJKKXITSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A selective inhibitor of peptidyl transferase center (PTC). In HL-60 (leukemia) cells, lycorine arrests the cell cycle at G2/M and induces apoptosis by a caspase-mediated pathway.

Caution

Protect from light.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjing Kang et al.
International immunopharmacology, 12(1), 249-256 (2011-12-14)
As a natural alkaloid extracted from Amaryllidaceae, lycorine shows various biological effects on tumor cells. Here we show that lycorine dose-dependently inhibited the LPS-induced up-regulation of nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) protein level in RAW264.7 cells. Besides, it
Carolyn Botting et al.
Expert opinion on drug discovery, 7(5), 417-428 (2012-03-24)
The members of the family Flaviviridae, including West Nile virus, yellow fever virus and dengue virus, are important human pathogens that are expanding their impact around the globe. The four serotypes of dengue infect 50-100 million people each year, yet
Ivan Ivanov et al.
Journal of plant physiology, 169(2), 206-211 (2011-11-29)
The alkaloid patterns in Leucojum aestivum L. shoot culture cultivated at temporary immersion conditions were investigated using gas chromatography-mass spectrometry. 18 alkaloids were identified, and galanthamine, hamayne and lycorine were dominant. The L. aestivum 80 shoot culture, cultivated at temporary
Yoshiaki Onishi et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 29(3-4), 407-416 (2012-04-18)
Plants of the Amaryllidaceae family have been used as therapeutic agents against CNS related maladies such as Alzheimer's disease. The known primary alkaloid constituents have significant biological activity. We identified the Lycoris alkaloids lycorine and lycoricidinol from Amaryllidaceae using a
Jiangning Liu et al.
Virology journal, 8, 483-483 (2011-10-28)
Human enterovirus 71 (EV71) infection causes hand, foot and mouth disease in children under 6 years old and this infection occasionally induces severe neurological complications. No vaccines or drugs are clinical available to control EV71 epidemics. In present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service