Skip to Content
Merck
All Photos(1)

Key Documents

L4170

Sigma-Aldrich

LDN-57444

≥98% (HPLC)

Synonym(s):

3-(O-acetyloxime), 5-chloro-1-[(2,5-dichlorophenyl)methyl]-1H-Indole-2,3-dione, C30, LDN 57444

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11Cl3N2O3
CAS Number:
Molecular Weight:
397.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated
protect from light

solubility

DMSO: 16 mg/mL

storage temp.

2-8°C

SMILES string

CC(=O)O\N=C1/C(=O)N(Cc2cc(Cl)ccc2Cl)c3ccc(Cl)cc13

InChI

1S/C17H11Cl3N2O3/c1-9(23)25-21-16-13-7-12(19)3-5-15(13)22(17(16)24)8-10-6-11(18)2-4-14(10)20/h2-7H,8H2,1H3/b21-16-

InChI key

OPQRFPHLZZPCCH-PGMHBOJBSA-N

Biochem/physiol Actions

LDN-57444 is a potent, reversible, competitive and active site-directed inhibitor of UCHL1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Beernaert et al.
European journal of cancer care, 25(4), 534-543 (2016-06-09)
Despite the growing consensus on the benefits of initiating palliative care early in the disease trajectory, it remains unclear at what point palliative care needs emerge. This study investigates quality of life and unmet palliative care needs at three phases
Giulia Chitarrini et al.
Protoplasma, 254(6), 2095-2106 (2017-03-23)
Metabolic changes can occur in plants in response to abiotic stress. Extensive use of leaf discs (mechanical wounding) in studies regarding the effect on the biochemical response of the grapevine to different types of biotic stress makes it necessary to
Chih-Wei Kao et al.
Plant molecular biology, 92(6), 643-659 (2016-11-01)
The beneficial root-colonizing fungus Piriformospora indica stimulates root development of Chinese cabbage (Brassica campestris subsp. Chinensis) and this is accompanied by the up-regulation of a τ-class glutathione (GSH)-S-transferase gene (BcGSTU) (Lee et al. 2011) in the roots. BcGSTU expression is
Thomas Vogl et al.
Nature communications, 9(1), 3589-3589 (2018-09-06)
Numerous synthetic biology endeavors require well-tuned co-expression of functional components for success. Classically, monodirectional promoters (MDPs) have been used for such applications, but MDPs are limited in terms of multi-gene co-expression capabilities. Consequently, there is a pressing need for new
Derrick Silove et al.
Social psychiatry and psychiatric epidemiology, 52(10), 1281-1294 (2017-08-22)
Cumulative evidence suggests that explosive anger may be a common reaction among survivors of mass conflict. However, little is known about the course of explosive anger in the years following mass conflict, or the psychosocial factors that influence the trajectory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service