Skip to Content
Merck
All Photos(1)

Key Documents

L1875

Sigma-Aldrich

L-(+)-Lactic acid solution

30% in H2O (by weight)

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720251
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

concentration

30% in H2O (by weight)

solubility

water: soluble(lit.)

storage temp.

2-8°C

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-(+)-Lactic acid solution may be used in the preparation of novel pH-sensitive physically crosslinked hydrogels. It may be used in the preparation of low molecular weight poly(lactic acid) by direct polycondensation.

Other Notes

Aqueous solutions of l(+)-Lactate.

Linkage

Purity equivalent to L 1750.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G G Jinesh et al.
Cell death and differentiation, 20(3), 382-395 (2012-11-24)
Caspases mediate apoptosis and have also been implicated in stem-cell biology. How caspases are linked to stem-cell biology is not known. Here, we show that the apoptotic blebs of cancer cells fuse together to form novel structures called 'blebbishields'. Blebbishields
Robert J Mishur et al.
Aging cell, 15(2), 336-348 (2016-01-06)
Disruption of mitochondrial respiration in the nematode Caenorhabditis elegans can extend lifespan. We previously showed that long-lived respiratory mutants generate elevated amounts of α-ketoacids. These compounds are structurally related to α-ketoglutarate, suggesting they may be biologically relevant. Here, we show
Debby Gawlitta et al.
Journal of applied physiology (Bethesda, Md. : 1985), 103(2), 464-473 (2007-04-21)
Prolonged periods of tissue compression may lead to the development of pressure ulcers, some of which may originate in, for example, skeletal muscle tissue and progress underneath intact skin, representing deep tissue injury. Their etiology is multifactorial and the interaction
Synthesis and characterization of pH-sensitive hydrogels based on chitosan and D, L-lactic acid.
Qu X, et al.
Journal of Applied Polymer Science, 74(13), 3193-3202 (1999)
Swelling and hydrolytic degradation of poly (D, L-lactic acid) in aqueous solutions.
Proikakis C S, et al.
Polymer Degradation and Stability, 91(3), 614-619 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service