Skip to Content
Merck
All Photos(1)

Key Documents

L0770

Sigma-Aldrich

Lectin from Pisum sativum (pea)

FITC conjugate, lyophilized powder

Synonym(s):

Pisum sativum agglutinin, PSA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

conjugate

FITC conjugate

Quality Level

form

lyophilized powder

potency

<20 μg per mL agglutination activity

composition

Protein, ≥95% biuret

extent of labeling

2-4 mol FITC per mol protein (Lowry).

impurities

salt, free

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin from Pisum sativum (pea) is a tetrameric glycoprotein of about 48 kDa.[1] It is isolated from peas.[1] Pisum sativum agglutinin is specific for α-D-glucose and α-D-mannose.[1]

Application

Lectin from Pisum sativum (pea) has been used to determine acrosomal status[2] and viability of spermatozoa[3] and to assess sperm acrosome reaction (AR) patterns[4]

Biochem/physiol Actions

Lectin from Pisum sativum (pea) possess anti-hepatoma effect and mitogenicity to lymphocytes, like concanavalin A (Con A).[5]
PSA is not blood group specific, but has an affinity for terminal α-D-mannosyl residues. PSA is a mitogen similar to concanavalin A.

Preparation Note

Conjugates are prepared from affinity purified lectin.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lectin-binding analysis of the biofilm exopolymeric matrix carbohydrate composition of corrosion-aggressive bacteria
Purish LM, et al.
Applied Biochemistry and Microbiology, 49(5), 458-463 (2013)
Martin Blomberg Jensen et al.
Nature communications, 12(1), 2450-2450 (2021-04-25)
Infertile men have few treatment options. Here, we demonstrate that the transmembrane receptor activator of NF-kB ligand (RANKL) signaling system is active in mouse and human testis. RANKL is highly expressed in Sertoli cells and signals through RANK, expressed in
Crypticity and functional distribution of the membrane associated ?-L-fucosidase of human sperm
Venditti JJ, et al.
Molecular Reproduction and Development, 74(6), 758-766 (2007)
Julio A Martinez-Rodriguez et al.
Reproduction & fertility, 1(1), 11-19 (2020-11-04)
Cryopreservation compromises the capacity of sperm fertilizing due to a series of alterations in the structure and physiology of the sperm. The use of antioxidants, such as melatonin, added to freezing media, may help to reduce sperm cryoinjury. To test
Amar K Chandra et al.
Environmental toxicology, 32(6), 1823-1835 (2017-02-17)
Excess iodine induced public health problems are now emerging in many iodine sufficient regions for indiscriminate intake of iodine through various iodized products. It has been reported that excess iodine can disrupt overall male reproductive physiology by generating oxidative stress

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service