Skip to Content
Merck
All Photos(1)

Key Documents

K3393

Sigma-Aldrich

PDK1, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PDPK1, PRO0461

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

102-138 nmol/min·mg

mol wt

~67 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDPK1(5170)

Biochem/physiol Actions

PDK1 (3-phosphoinositide-dependent protein kinase) is activated by the presence of PtdIns (3, 4, 5) P3 or PtdIns (3, 4) P2. PDK1 then activates protein kinase B (PKB) which, in turn, inactivates glycogen synthase kinase-3 (GSK3). The phosphorylation of other proteins by PKB and GSK3 is likely to mediate many of the intracellular actions of insulin. Thus, PDK1 plays a key role in mediating many of the actions of the second messenger(s) PtdIns (3, 4, 5) P3 and/or PtdIns (3, 4) P2. The human PDK1 is a 556-residue monomeric enzyme comprising of a catalytic domain that is most similar to the PKA, PKB and PKC subfamily of protein kinases.
PDPK1 (3-phosphoinositide dependent protein kinase 1) is a serine/threonine protein kinase which is activated upon accumulation of PIP3 (phosphatidylinositol-3,4,5-trisphosphate), a PI3K (phosphoinositide 3-kinase) product. It controls AGC (protein kinase A, G, and C) kinase members, such as AKT (v-akt murine thymoma viral oncogene homolog), p70 ribosomal S6 kinase (S6K), serum- and glucocorticoid-induced protein kinase (SGK), and protein kinase C (PKC) members. It is involved in metabolism, growth, proliferation, and survival. PDPK1 is associated with growth of angiosarcoma cells.

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Zabkiewicz et al.
Haematologica, 99(5), 858-864 (2013-12-18)
PDK1 is a master kinase that activates at least six protein kinase groups including AKT, PKC and S6K and is a potential target in the treatment of a range of malignancies. Here we show overexpression of PDK1 in over 40%
Jing Tan et al.
Cancer discovery, 3(10), 1156-1171 (2013-07-28)
Although 3-phosphoinositide-dependent protein kinase-1 (PDK1) has been predominately linked to the phosphoinositide 3-kinase (PI3K)-AKT pathway, it may also evoke additional signaling outputs to promote tumorigenesis. Here, we report that PDK1 directly induces phosphorylation of Polo-like kinase 1 (PLK1), which in
Makoto Wada et al.
Journal of dermatological science, 78(1), 44-50 (2015-03-03)
Angiosarcoma is a rare and aggressive malignant neoplasm of endothelial cells. Recent studies have shown that the mTOR pathway is also aberrantly activated in cutaneous angiosarcoma. New therapeutic strategies are required because the prognosis of this disease is still poor.
D R Alessi et al.
Current biology : CB, 7(4), 261-269 (1997-04-01)
Protein kinase B (PKB), also known as c-Akt, is activated rapidly when mammalian cells are stimulated with insulin and growth factors, and much of the current interest in this enzyme stems from the observation that it lies 'downstream' of phosphoinositide
P Cohen et al.
FEBS letters, 410(1), 3-10 (1997-06-23)
The initial steps in insulin signal transduction occur at the plasma membrane and lead to the activation of phosphatidylinositide (PtdIns) 3-kinase and the formation of PtdIns(3,4,5,)P3 in the inner leaflet of the plasma membrane which is then converted to PtdIns(3,4)P2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service