Skip to Content
Merck
All Photos(2)

Key Documents

I8636

Sigma-Aldrich

D-myo-Inositol 1,3,4,5-tetrakis(phosphate) ammonium salt

≥95%

Synonym(s):

IP4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H16O18P4 · 8NH3
Molecular Weight:
636.32
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95%

storage temp.

−20°C

SMILES string

N.O[C@H]1[C@H](OP(O)(O)=O)[C@H](O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O

InChI

1S/C6H16O18P4.H3N/c7-1-3(21-25(9,10)11)2(8)5(23-27(15,16)17)6(24-28(18,19)20)4(1)22-26(12,13)14;/h1-8H,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20);1H3/t1-,2-,3-,4+,5-,6-;/m0./s1

InChI key

LDRGGXGJARVUBP-SUBVTSALSA-N

Biochem/physiol Actions

Precursor of inositol 1,3,4-trisphosphate; increases entry of calcium across the plasma membrane; proposed regulator of intracellular calcium by control of plasma membrane transport of calcium ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonghui Jia et al.
Cell cycle (Georgetown, Tex.), 7(18), 2803-2808 (2008-09-05)
Many neutrophil functions are mediated by PtdIns(3,4,5)P3 that exerts its role by mediating protein translocation via binding to their PH-domains. Inositol 1,3,4,5-tetrakisphosphate (Ins(1,3,4,5)P4) binds the same PH domain, competes for its binding to PtdIns(3,4,5)P3, and thus negatively regulates PtdIns(3,4,5)P3 signaling.
Karsten Sauer et al.
Nature reviews. Immunology, 10(4), 257-271 (2010-03-26)
The membrane lipid phosphatidylinositol-3,4,5-trisphosphate (PtdInsP(3)) regulates membrane receptor signalling in many cells, including immunoreceptor signalling. Here, we review recent data that have indicated essential roles for the soluble PtdInsP(3) analogue inositol-1,3,4,5-tetrakisphosphate (InsP(4)) in T cell, B cell and neutrophil development
Reiko Sakaguchi et al.
Bioorganic & medicinal chemistry, 17(20), 7381-7386 (2009-09-19)
A fluorescent sensor for the detection of inositol-1,3,4,5-tetrakisphosphate, Ins(1,3,4,5)P(4), was constructed from a split PH domain and a single circularly permuted GFP. A structure-based design was conducted to transduce a ligand-induced subtle structural perturbation of the split PH domain to
Xavier Pesesse et al.
Biochemical and biophysical research communications, 336(1), 157-162 (2005-09-06)
Hyperosmotic stress induces water diffusion out of the cell, resulting in cell shrinkage, and leading to DNA damage, cell cycle arrest, and cytoskeletal reorganization. A previous report showed that low concentrations of sorbitol (200mM) could increase up to 25-fold the
Natalia Riquelme et al.
Foods (Basel, Switzerland), 9(1) (2020-01-18)
The aim of this work was to study the effect of two emulsifiers (M1: SL-soy lecithin, Tw80-Tween 80 and CasCa-calcium caseinate and M2: SL-soy lecithin, Tw80-Tween 80 and SE-sucrose esters) on the oxidative stability of avocado oil-based nanoemulsions. Oil-in-water nanoemulsions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service