Skip to Content
Merck
All Photos(1)

Key Documents

I1911

Sigma-Aldrich

IU1

≥98% (HPLC)

Synonym(s):

1-[1-(4-Fluoro-phenyl)-2,5-dimethyl-1H-pyrrol-3-yl]-2-pyrrolidin-1-yl-ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21FN2O
CAS Number:
Molecular Weight:
300.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

FC1=CC=C(N2C(C)=CC(C(CN3CCCC3)=O)=C2C)C=C1

InChI

1S/C18H21FN2O/c1-13-11-17(18(22)12-20-9-3-4-10-20)14(2)21(13)16-7-5-15(19)6-8-16/h5-8,11H,3-4,9-10,12H2,1-2H3

InChI key

JUWDSDKJBMFLHE-UHFFFAOYSA-N

Application

IU1 has been used for the inhibition of Ubiquitin Specific Peptidase 14 (USP14) in human neuroblastoma cells (SH-SY5Y) and in ubiquitin-rhodamine hydrolysis plate assay.

Biochem/physiol Actions

IU1 is an inhibitor of USP14, a deubiquitinating enzyme associated with the proteasome. The proteasome mediates the cellular degradation of oxidized, damaged and misfolded proteins which, if not removed, accumulate and become toxic to cells. Proteins targeted for proteasomal degradation are first ubiquitinated, with longer length ubiquitin chains interacting more strongly with the proteasome. Deubiquitinating enzymes (DUBs) such as USP14 interfere with the degradation process. IU1 inhibits USP14-mediated ubiquitin "chain-trimming" thereby enhancing substrate degradation by the proteasome. The compound may help to eliminate toxic proteins more effectively by enhancing their degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ningning Liu et al.
Molecular and cellular biochemistry, 431(1-2), 87-96 (2017-04-02)
Persistent activation of nuclear factor B (NF-κB) is very important in the modulation of macrophages cellular response to microbial infections. The deubiquitinase USP14, which is critical for ubiquitin-mediated proteasomal degradation of proteins, is known to be involved in cancer, neurological
USP14 inhibition corrects an in vivo model of impaired mitophagy.
Chakraborty J, et al.
EMBO Molecular Medicine, 10(11), e9014-e9014 (2018)
Liu Xu et al.
International journal of biological sciences, 16(15), 2951-2963 (2020-10-17)
Previous studies have demonstrated that the antitumor potential of IU1 (a pharmacological compound), which was mediated by selective inhibition of proteasome-associated deubiquitinase ubiquitin-specific protease 14 (USP14). However, the underlying molecular mechanisms remain elusive. It has been well established that mdm2
Inactive USP14 and inactive UCHL5 cause accumulation of distinct ubiquitinated proteins in mammalian cells.
Chadchankar J, et al.
bioRxiv, 10(11), 479758-479758 (2018)
Yuning Liao et al.
Cell death & disease, 8(2), e2585-e2585 (2017-02-06)
Androgen receptor (AR) is frequently over-expressed and plays a critical role in the growth and progression of human prostate cancer. The therapy attempting to target AR signalling was established in decades ago but the treatment of prostate cancer is far

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service