Recommended Products
recombinant
expressed in E. coli
Quality Level
packaging
pkg of (6 vials per kit)
application(s)
CRISPR
genome editing
selection
kanamycin
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
The CRISPR Integration Kit provides the essential genome editing reagents necessary to integrate a BstNI restriction site to the human KRAS locus. This kit is intended to serve as a tool to evaluate cell lines for ability to integrate new sequences at CRISPR-effected double strand breaks with the aid of short oligonucleotide donor. Integration efficiencies obtained with this kit enable rational design of genome editing experiments in a wide range of cell lines.
Application
CRISPR/Cas9 can be used to introduce, or “knock in”, new DNA sequences. Common modifications include the introduction of a SNP, small tag, loxP or larger cassette such as fluorescent protein. These modifications are made through the introduction of targeted DSB which enhance targeted integration significantly (Choulika et al., 1995). Targeted integration (gene knock in) occurs through homology directed repair (HDR)( (Bibikova et al., 2002). In order to enable gene editing by HDR, a DNA ‘donor′ or repair template containing the desired sequence must be delivered to the cell along with gRNA and Cas9.
Features and Benefits
- The CRISPR Integration Assay provides a validated CRISPR that cleaves DNA with high efficiency along with a standardized oligo donor designed to survey homology dependent repair frequencies in a wide variety of cell lines
- CRISPR-aided integration increases the targeted mutagenesis rate without the need for antibiotic selection
- Integration efficiencies estimated with this kit enable rational design of genome editing experiments in a wide range of cell lines
Components
Each kit consists of:
one vial of gRNA plasmid - KRAS
one vial of Cas9 GFP plasmid
one vial of 120 base donor oligonucleotide
one vial of forward PCR primer
one vial of reverse PCR primer
one vial of genomic DNA control
one vial of gRNA plasmid - KRAS
one vial of Cas9 GFP plasmid
one vial of 120 base donor oligonucleotide
one vial of forward PCR primer
one vial of reverse PCR primer
one vial of genomic DNA control
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service