HS23471B
ClearlySafe Glove Box Holder
Fits 2 glove boxes, clear acrylic
Synonym(s):
glove dispenser
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
material
clear acrylic
feature
Holds 2 boxes
manufacturer/tradename
Heathrow Scientific HS23471B
W × H × D
295 × 260 × 110 mm
Related Categories
Features and Benefits
- All holders fit most of the box brands
- Mounting screws are provided
- A polyester foam insert is included in the clear acrylic holders to firmly hold a variety of glove box sizes-snug fit facilitates quick removal of gloves.
- all holders fit most major brands of boxes
- mounting screws included
- includes a polyester foam insert to securely hold a range of glove sizes- snug fit ensures easy removal of gloves
Other Notes
Gloves to be organized and kept ready for immediate use. Free up benchtop space by mounting holders on the wall for convenient glove dispensing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)