HS15994P
Premium slide box
100 place, foam, red plastic
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
All Photos(1)
About This Item
UNSPSC Code:
41123302
NACRES:
NB.17
Recommended Products
material
nickel closure
red plastic
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994P
L × W × H
208 × 175 × 34 mm
Features and Benefits
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism