HS15989C
Premium slide box
25 place, cork lining, red plastic
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
All Photos(1)
About This Item
UNSPSC Code:
41123302
NACRES:
NB.17
Recommended Products
material
nickel closure
red plastic
feature
Holds 25 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15989C
L × W × H
141 × 88 × 35 mm
Related Categories
Features and Benefits
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism
Choose from one of the most recent versions:
Certificates of Analysis (COA)