HS15981A
Slide holder
for 40 slides, white polyethylene
All Photos(1)
About This Item
UNSPSC Code:
41123302
NACRES:
NB.35
Recommended Products
material
white polyethylene
feature
Holds 40 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15981A
size
298x105x24 mm
Related Categories
General description
The Slide Holder holds slides during evaluation, cleaning, and drying.
- Made from chemical-resistant polyethylene
- Solid, strong construction with rubber feet for bench top stability
Choose from one of the most recent versions:
Certificates of Analysis (COA)