HS120226
Pour Boat Weighing Dishes
medium, white polystyrene (Anti-static), pk of 500
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
All Photos(2)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Recommended Products
material
white polystyrene (Anti-static)
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific HS120226
L × W × H
83 mm × 132 mm × 26 mm
Related Categories
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- pour-spout design easily dispenses samples without flexing the dish
- flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling
Choose from one of the most recent versions:
Certificates of Analysis (COA)