Skip to Content
Merck
All Photos(1)

Key Documents

HPA043264

Sigma-Aldrich

Anti-HSD3B1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSD3B, Anti-HSDB3, Anti-SDR11E1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RHIVTLSNSVFTFSYKKAQRDLAYKPLYSWEEAKQKT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD3B1(3283)

General description

3 β-hydroxysteroid dehydrogenase/delta 5->4-isomerase type 1 (HSD3B1) is a key enzyme essential for the formation of steroid sex hormones. It is majorly expressed in mammary glands, prostate, placenta and sebaceous glands of skin. HSD3B1 gene is mapped to human chromosome 1p12 and has four exons.

Immunogen

hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

3 β-hydroxysteroid dehydrogenase/delta 5->4-isomerase type 1 (HSD3B1) catalyses the conversion of steroid dehydroepiandrosterone sulfate to estrogen precursors hormones. It regulates levels of dihydrotestosterone (DHT). HSD3B1 is highly expressed in breast cancer and HSD3B1 gene knockdown results in tumor growth inhibition. A single nucleotide polymorphism in the HSD3B1 gene enhances enzyme activity and improves dihydrotestosterone (DHT) production in castration-resistant prostate cancer (CRPC). Expression of HSD3B1 is downregulated in ovarian cancer. Polymorphism in HSD3B1 is implicated in individuals with hypertension.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82530

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphism in exon 4 of the human 3beta-hydroxysteroid dehydrogenase type I gene (HSD3B1) and blood pressure
Rosmond R, et al.
Biochemical and Biophysical Research Communications, 293(1), 629-632 (2002)
Metabolism of dihydrotestosterone in human liver: importance of 3alpha-and 3beta hydroxysteroid dehydrogenase
Pirog EC and Collins DC
The Journal of Clinical Endocrinology and Metabolism, 84(9), 3217-3221 (1999)
Human 3beta-hydroxysteroid dehydrogenase type 1 in human breast cancer: clinical significance and prognostic associations
Hanamura T, et al.
Cancer Medicine, 5(7), 1405-1415 (2016)
Variant allele of HSD3B1 increases progression to castration-resistant prostate cancer
Wu G, et al.
Prostate, 75(7), 777-782 (2015)
Expression of 3beta-hydroxysteroid dehydrogenase type 1 in breast cancer is associated with poor prognosis independent of estrogen receptor status
Chang Y, et al.
Annals of Surgical Oncology, 24(13), 4033-4041 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service