Skip to Content
Merck
All Photos(6)

Key Documents

HPA040376

Sigma-Aldrich

Anti-TOX3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cagf9, Anti-Tnrc9, Anti-Tox high mobility group box family member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SQGSEFTPQFPPQSLDLPSITISRNLVEQDGVLHSSGLHMDQSHTQVSQYRQDPSLIMRSIVHMTDAARSGVMP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOX3(27324)

General description

The TOX high mobility group box family member 3 (TOX3) gene with seven exons, is mapped to human chromosome 16q12. The encoded protein is mainly expressed in brain and breast. TOX3 includes a high mobility group box (HMG-box) domain and a C- terminal polyglutamine stretch.

Immunogen

TOX high mobility group box family member 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-TOX3 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

TOX high mobility group box family member 3 (TOX3) interacts with cAMP response element (CRE)-binding protein (CREB) and CREB-binding protein (CBP) and regulates calcium-dependent transcription in neurons. Overexpression or mutation of the gene is associated with the development and progression of breast cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81832

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Owain Jones et al.
PloS one, 8(9), e74102-e74102 (2013-09-27)
TOX3 maps to 16q12, a region commonly lost in breast cancers and recently implicated in the risk of developing breast cancer. However, not much is known of the role of TOX3 itself in breast cancer biology. This is the first
Shauna H Yuan et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(8), 2909-2914 (2009-02-07)
We report the cloning and characterization of TOX3, a high mobility group box protein involved in mediating calcium-dependent transcription. TOX3 was identified as a calcium-dependent transactivator using the Transactivator Trap screen. We find that TOX3 interacts with both cAMP response
Sanjeeb Kumar Sahu et al.
Biochimica et biophysica acta, 1859(7), 833-840 (2016-04-16)
The human genomic locus for the transcription factor TOX3 has been implicated in susceptibility to restless legs syndrome and breast cancer in genome-wide association studies, but the physiological role of TOX3 remains largely unknown. We found Tox3 to be predominantly
Cui-Cui Han et al.
Oncology letters, 11(3), 1762-1768 (2016-03-22)
TOX3 is a newly identified gene that has been observed to correlate with breast cancer by genome-wide association studies (GWAS) in recent years. In addition, it has been noted that single-nucleotide polymorphisms (SNPs) in the TOX3 gene have a strong
Helmuth Gehart et al.
Cell, 176(5), 1158-1173 (2019-02-05)
Homeostatic regulation of the intestinal enteroendocrine lineage hierarchy is a poorly understood process. We resolved transcriptional changes during enteroendocrine differentiation in real time at single-cell level using a novel knockin allele of Neurog3, the master regulator gene briefly expressed at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service