Skip to Content
Merck
All Photos(7)

Key Documents

HPA036362

Sigma-Aldrich

Anti-LACTB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ14902, Anti-Lactamase, β, Anti-MRPL56

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KEGKSNEKNDFTKFKTEQENEAKCRNSKPGKKKNDFEQGELYLREKFENSIESLRLFKNDPLFFKPGSQFLYSTFGYTLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LACTB(114294)

General description

Lactamase beta (LACTB) is a 60kDa mammalian mitochondrial protein. The protein shows sequence similarity to sequence β-lactamases and penicillin binding proteins (PBPs), which occurs in bacteria. It is expressed in skeletal muscle, heart and liver. The gene is located on human chromosome 15q22.2.

Immunogen

lactamase, beta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Lactamase beta (LACTB) inhibits the proliferation of breast cancer cells, by modifying the mitochondrial lipid metabolism and differentiation of breast cancer cells. It contributes to intra-mitochondrial membrane organization, modulates complex I of the mitochondrial electron transport chain and controls cellular metabolic processes. This protein might act as a therapeutic target for gliomas. It may function as a tumor suppressor gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78751

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LACTB is a tumour suppressor that modulates lipid metabolism and cell state
Keckesova Z, et al.
Nature, 543(7647), 681-681 (2017)
Genome-wide association study on detailed profiles of smoking behavior and nicotine dependence in a twin sample.
Loukola A
Molecular Psychiatry, 19(5), 615-624 (2014)
Expression and purification of the mitochondrial serine protease LACTB as an N-terminal GST fusion protein in Escherichia coli
Liobikas J, et al.
Protein Expression and Purification, 45(2), 335-342 (2006)
Overexpression of LACTB, a Mitochondrial Protein, that Inhibits Proliferation and Invasion in Glioma Cells.
Li HT, et al.
Oncology Research, 335-342 (2017)
LACTB-mediated tumour suppression by increased mitochondrial lipid metabolism
Cucchi D and Mauro C
Cell Death and Differentiation, 24, 335-342 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service