Skip to Content
Merck
All Photos(7)

Key Documents

HPA035981

Sigma-Aldrich

Anti-PDCD1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD279, Anti-PD1, Anti-SLEB2, Anti-hSLE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

WFLDSPDRPWNPPTFSPALLVVTEGDNATFTCSFSNTSESFVLNWYRMSPSNQTDKLAAFPEDRSQPGQDCRFRVTQLPNGRDFHMSVVRARRNDSGTYLCGAIS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD1(5133)

General description

The gene PDCD1 (programmed cell death protein 1) is mapped to human chromosome 2q37. The encoded protein belongs to the CD28 superfamily. It interacts with its ligands PD-L1/PD-L2 (programmed death-ligands).

Immunogen

programmed cell death 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-PDCD1 antibody produced in rabbit has been used for immunohistochemistry.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PDCD1 (programmed cell death protein 1) plays an important role in the immune regulation by working as a negative co-stimulatory factor. It suppresses working and multiplication of the T cells, thereby reducing the release of IL-2 (interleukin-2), IL-10 and IFN-γ (interferon γ). In the presence of tumor microenvironment, PDCD1 along with its ligand modulates T effector functioning, thereby protecting cancer cells from immune-associated rejection. Mutation in this gene is associated with symptoms and progression of multiple sclerosis. Mutation in it are also linked with susceptibility to systemic lupus erythematosus.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70027

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elevated Cellular PD1/PD-L1 Expression Confers Acquired Resistance to Cisplatin in Small Cell Lung Cancer Cells.
Yan F
PLoS ONE, 11(9) (2016)
The Expression of Programmed Death-1 on CD4+ and CD8+ T Lymphocytes in Patients with Type 2 Diabetes and Severe Sepsis.
Jia Y
PLoS ONE, 11(7) (2016)
Zijun Y Xu-Monette et al.
Blood, 131(1), 68-83 (2017-11-10)
Programmed cell death protein 1 (PD-1) blockade targeting the PD-1 immune checkpoint has demonstrated unprecedented clinical efficacy in the treatment of advanced cancers including hematologic malignancies. This article reviews the landscape of PD-1/programmed death-ligand 1 (PD-L1) expression and current PD-1
Mengdi He et al.
Journal of gynecologic oncology, 32(3), e32-e32 (2021-04-08)
To predict the prognosis of cervical cancer, we constructed a novel model with 5 specific cell types and identified a potential biomarker. We employed CIBERSORT and xCell method to evaluate the abundances of 23 cells types in tumor microenvironment. Five
A regulatory polymorphism in PDCD1 is associated with susceptibility to systemic lupus erythematosus in humans.
Prokunina L
Nature Genetics, 32(4), 666-669 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service