Skip to Content
Merck
All Photos(2)

Key Documents

HPA029856

Sigma-Aldrich

Anti-LALBA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LYZL7, Anti-Lactalbumin, α-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

QVPQSRNICDISCDKFLDDDITDDIMCAKKILDIKGIDYWLAHKALCTEKLEQWLCEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LALBA(3906)

General description

The gene LALBA (α-lactalbumin) is mapped to human chromosome 12q13. It is a small acidic calcium-binding milk protein. LALBA constitutes about 20-25% of total protein in human milk. The protein has a helical α-domain and a β-sheeted β-domain.

Immunogen

lactalbumin, alpha- recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-LALBA antibody produced in rabbit has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

LALBA (α-lactalbumin) is part of the lactose synthase complex and is responsible for the biosynthesis of lactose. Peptides generated after partial digestion of this protein have antimicrobial and immunostimulatory properties. A multimeric α-lactalbumin enhances cell death in tumors.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77903

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent K Tuohy et al.
Cancers, 8(6) (2016-06-21)
We have proposed that safe and effective protection against the development of adult onset cancers may be achieved by vaccination against tissue-specific self-proteins that are "retired" from expression at immunogenic levels in normal tissues as we age, but are overexpressed
The oleic acid complexes of proteolytic fragments of alpha-lactalbumin display apoptotic activity.
Tolin S, et al.
FEBS Journal, 277, 163-173 (2010)
The human alpha-lactalbumin molten globule: comparison of structural preferences at pH 2 and pH 7.
Rosner HI and Redfield C
Journal of Molecular Biology, 394, 351-362 (2009)
The protective role of pregnancy in breast cancer.
Russo J, et al.
Breast Cancer Research, 7, 131-142 (2005)
Targeted Vaccination against Human a-Lactalbumin for Immunotherapy and Primary Immunoprevention of Triple Negative Breast Cancer.
Tuohy VK, et al.
Cancers, 8 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service